missing translation for 'onlineSavingsMsg'
Learn More

(3-Aminopropyl)triethoxysilane, 98%

Alfa Aesar (3-Aminopropyl)triethoxysilane, 98%, Quantity: 100g, Boiling Point: 217 deg.C, CAS: 919-30-2, Molecular Formula: C9H23NO3Si, Molecular Weight: 221.372, Percent Purity: 98%, pH: 11 | CAS: 919-30-2 | C9H23NO3Si | 221.37 g/mol

$103.27 - $969.15

Chemical Identifiers

CAS 919-30-2
Molecular Formula C9H23NO3Si
Molecular Weight (g/mol) 221.37
MDL Number MFCD00008207,MFCD01324904
InChI Key WYTZZXDRDKSJID-UHFFFAOYSA-N
Synonym 3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane
PubChem CID 13521
IUPAC Name 3-triethoxysilylpropan-1-amine
SMILES CCO[Si](CCCN)(OCC)OCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1066822
View Documents
Thermo Scientific Chemicals
A1066822
100 g
Each for $103.27
Only null left
 
AAA1066836
View Documents
Thermo Scientific Chemicals
A1066836
500 g
Each for $248.90
Only null left
 
AAA106680E
View Documents
Thermo Scientific Chemicals
A106680E
2500 g
Each for $969.15
Only null left
 
Description

Description

(3-Aminopropyl)triethoxysilane is useful in the formation of aminopropyl derivative of glass. It acts as an adsorbent for affinity chromatography. It is used to prepare positively charged slides suitable for use with various immunohistochemical and in situ hybridization procedures. In addition, it is used as adhesives and sealant chemicals, paint additives and coating additives. Further, it is used as a silylation reagent for coating glass and silica surfaces and to crosslink and immobilize proteins and other molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Aminopropyl)triethoxysilane is useful in the formation of aminopropyl derivative of glass. It acts as an adsorbent for affinity chromatography. It is used to prepare positively charged slides suitable for use with various immunohistochemical and in situ hybridization procedures. In addition, it is used as adhesives and sealant chemicals, paint additives and coating additives. Further, it is used as a silylation reagent for coating glass and silica surfaces and to crosslink and immobilize proteins and other molecules.

Solubility
Miscible with toluene, acetone, chloroform and ethanol.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, acids and moisture.
Specifications

Chemical Identifiers

919-30-2
221.37
WYTZZXDRDKSJID-UHFFFAOYSA-N
13521
CCO[Si](CCCN)(OCC)OCC
C9H23NO3Si
MFCD00008207,MFCD01324904
3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane
3-triethoxysilylpropan-1-amine

Specifications

217°C
11
C9H23NO3Si
MFCD00008207,MFCD01324904
1754988
3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane
WYTZZXDRDKSJID-UHFFFAOYSA-N
3-triethoxysilylpropan-1-amine
13521
98%
1.421
0.948
919-30-2
100 g
H2N(CH2)3Si(OCH2CH3)3
UN2735
104°C (219°F)
Miscible with toluene,acetone,chloroform and ethanol.
CCO[Si](CCCN)(OCC)OCC
221.37
221.37
Amine-like
Air and moisture sensitive
(3-Aminopropyl)triethoxysilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.

P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H302-H314-H317-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 213-048-4

RTECSNumber : TX2100000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

RUO – Research Use Only