missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3-Aminopropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A112840E

 View more versions of this product

Catalog No. AAA112840E


Explore available promotions

Description

Description

(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).

Solubility
Miscible with water.

Notes
Moisture sensitive. Incompatible withstrong acids, strong oxidizing agents, water, bases, alcohols and peroxides.
Specifications

Specifications

(3-Aminopropyl)trimethoxysilane
1.02
90°C (194°F)
C6H17NO3Si
H2N(CH2)3Si(OCH3)3
2500 g
2038118
3-aminopropyltrimethoxysilane, 3-aminopropyl trimethoxysilane, 3-trimethoxysilyl-1-propanamine, 3-trimethoxysilyl propan-1-amine, 3-trimethoxysilyl propylamine, 1-propanamine, 3-trimethoxysilyl, silane sc 3900, n-trimethoxysilylpropyl amine, propylamine, 3-trimethoxysilyl, unii-7mry0c9sb6
SJECZPVISLOESU-UHFFFAOYSA-N
3-trimethoxysilylpropan-1-amine
83756
97%
13822-56-5
194°C
Amine-like
1.424
MFCD00008206
UN2735
Air and moisture sensitive
Miscible with water.
CO[Si](CCCN)(OC)OC
179.291
179.29

Chemical Identifiers

13822-56-5
179.291
SJECZPVISLOESU-UHFFFAOYSA-N
83756
CO[Si](CCCN)(OC)OC
C6H17NO3Si
MFCD00008206
3-aminopropyltrimethoxysilane, 3-aminopropyl trimethoxysilane, 3-trimethoxysilyl-1-propanamine, 3-trimethoxysilyl propan-1-amine, 3-trimethoxysilyl propylamine, 1-propanamine, 3-trimethoxysilyl, silane sc 3900, n-trimethoxysilylpropyl amine, propylamine, 3-trimethoxysilyl, unii-7mry0c9sb6
3-trimethoxysilylpropan-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P370+P378q-P501c

H227-H315-H318

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 237-511-5

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only