Learn More
3-Aminopropyltriethoxysilane, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 151080050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Aminopropyltriethoxysilane | |
-70.0°C | |
217.0°C | |
Authentic | |
Glass bottle | |
1.4200 to 1.4220 | |
MFCD00008207,MFCD01324904 | |
04,IV,4273 | |
3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane | |
WYTZZXDRDKSJID-UHFFFAOYSA-N | |
3-triethoxysilylpropan-1-amine | |
13521 | |
99% |
919-30-2 | |
0.9420g/mL | |
96°C | |
98.5% min. (GC) | |
C9H23NO3Si | |
H2N(CH2)3Si(OC2H5)3 | |
5 g | |
0.942 | |
Solubility in water: reacts. Other solubilities: soluble in chloroform | |
CCO[Si](CCCN)(OCC)OCC | |
221.37 | |
221.37 | |
Liquid |
Chemical Identifiers
919-30-2 | |
221.37 | |
WYTZZXDRDKSJID-UHFFFAOYSA-N | |
13521 | |
CCO[Si](CCCN)(OCC)OCC |
C9H23NO3Si | |
MFCD00008207,MFCD01324904 | |
3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane | |
3-triethoxysilylpropan-1-amine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 213-048-4
RUO – Research Use Only