Learn More
3-Aminopyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals
$278.45 - $700.78
Chemical Identifiers
CAS | 1462-86-8 |
---|---|
Molecular Formula | C6H6N2O2 |
Molecular Weight (g/mol) | 138.13 |
MDL Number | MFCD00090153 |
InChI Key | BOOMHTFCWOJWFO-UHFFFAOYSA-N |
Synonym | 3-aminopicolinic acid, 3-amino-2-pyridinecarboxylic acid, 3-amino-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 3-amino, 3-amino-2-picolinic acid, 3-amino-2-pyridinecarboxylicacid, 3-amino-2-carboxypyridine, 2-pyridinecarboxylicacid, 3-amino, pubchem2015, 3-aminopyridine-2-carboxylic acid, tech. |
PubChem CID | 73836 |
IUPAC Name | 3-aminopyridine-2-carboxylic acid |
SMILES | C1=CC(=C(N=C1)C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC432520050
|
Thermo Scientific Chemicals
432520050 |
5 g | Glass bottle |
Each for $278.45
|
|
|||||
AC432520250
|
Thermo Scientific Chemicals
432520250 |
25 g | Glass bottle |
Each for $700.78
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1462-86-8 | |
138.13 | |
BOOMHTFCWOJWFO-UHFFFAOYSA-N | |
73836 | |
C1=CC(=C(N=C1)C(=O)O)N |
C6H6N2O2 | |
MFCD00090153 | |
3-aminopicolinic acid, 3-amino-2-pyridinecarboxylic acid, 3-amino-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 3-amino, 3-amino-2-picolinic acid, 3-amino-2-pyridinecarboxylicacid, 3-amino-2-carboxypyridine, 2-pyridinecarboxylicacid, 3-amino, pubchem2015, 3-aminopyridine-2-carboxylic acid, tech. | |
3-aminopyridine-2-carboxylic acid |
Specifications
1462-86-8 | |
100.0 | |
Brown to Yellow | |
96% min. (HPLC) | |
C6H6N2O2 | |
5 g | |
BOOMHTFCWOJWFO-UHFFFAOYSA-N | |
3-aminopyridine-2-carboxylic acid | |
73836 | |
97% | |
3-Aminopyridine-2-carboxylic acid |
96.0 | |
200°C to 201°C | |
Authentic | |
Glass bottle | |
MFCD00090153 | |
3-aminopicolinic acid, 3-amino-2-pyridinecarboxylic acid, 3-amino-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 3-amino, 3-amino-2-picolinic acid, 3-amino-2-pyridinecarboxylicacid, 3-amino-2-carboxypyridine, 2-pyridinecarboxylicacid, 3-amino, pubchem2015, 3-aminopyridine-2-carboxylic acid, tech. | |
C1=CC(=C(N=C1)C(=O)O)N | |
138.13 | |
138.13 | |
Crystalline Powder or Powder |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 215-971-8
RUO – Research Use Only