missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Aminoquinuclidine dihydrochloride, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 146150250

Catalog No. AC146150250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Aminoquinuclidine dihydrochloride
6530-09-2
100.0
98%
C7H14N2·2ClH
25g
Solubility in water: soluble
[H+].[H+].[Cl-].[Cl-].NC1CN2CCC1CC2
199.12
CHEBI:47953
98%
98%
97.5
Authentic
Glass bottle
MFCD00137395
2s,3s,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexanoic acid, aldehydo-d-glucuronic acid, unii-8a5d83q4rw, rel, d-glucuronic acid, d-+ glucuronic acid, d03jeu, d-glucuronic acid 5g
STZHBULOYDCZET-UHFFFAOYNA-N
(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
65041
199.12
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 229-424-6

RTECSNumber : VD4590000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only