missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals 3'-Azido-3'-deoxythymidine, 98%

Anti-viral agent

Supplier:  Thermo Scientific Chemicals J6512706

 View more versions of this product

Catalog No. AAJ6512706


Explore available promotions

Description

Description

3'-Azido-3'-deoxythymidine is a Anti-viral agent, and also used as a reverse transcriptase inhibitor active against HIV-1 virus that blocks the incorporation of nucleotides into growing DNA strands. Useful for antiviral and anticancer studies. 3?-Azido-3?-deoxythymidine enters CNS by diffusion and causes irreversible telomere shortening.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′-Azido-3′-deoxythymidine is a Anti-viral agent, and also used as a reverse transcriptase inhibitor active against HIV-1 virus that blocks the incorporation of nucleotides into growing DNA strands. Useful for antiviral and anticancer studies. 3′-Azido-3′-deoxythymidine enters CNS by diffusion and causes irreversible telomere shortening.

Solubility
Soluble in water.

Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

3'-Azido-3'-deoxythymidine
113°C to 115°C
C10H13N5O4
5 g
Light sensitive; Very Hygroscopic
azt; azidothymidine; zdv; zidovudine, 1-2r-4-azido-5-hydroxymethyl oxolan-2-yl-5-methyl-3h-pyrimidine-2,4-dione
HBOMLICNUCNMMY-CFQLRCIDNA-N
1-[(2R,4S,5S)-4-azido-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
455007
98%
30516-87-1
White
MFCD00006536
3595791
1410123
Soluble in water.
CC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](CO)O2)C(=O)NC1=O
267.25
267.24
Powder

Chemical Identifiers

30516-87-1
267.25
HBOMLICNUCNMMY-CFQLRCIDNA-N
455007
C10H13N5O4
MFCD00006536
azt; azidothymidine; zdv; zidovudine, 1-2r-4-azido-5-hydroxymethyl oxolan-2-yl-5-methyl-3h-pyrimidine-2,4-dione
CC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](CO)O2)C(=O)NC1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes serious eye irritation, Suspected of causing genetic defects, May cause respiratory irritation, Suspected of causing cancer, Causes skin irritation

Serious eye damage/eye irritation (category 2), Germ cell mutagenicity (category 2), Specific target organ toxicity after single exposure (category 3), Carcinogenicity (category 2), Skin corrosion/irritation (category 2)

RTECSNumber : XP207200

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only