missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Azidopropylamine 95.0+%, TCI America™

Supplier:  TCI America A2738100MG

Catalog No. A2738100MG


Explore available promotions

Description

Description

Specifications

Specifications

3-Azidopropylamine
Yellow
C3H8N4
100 mg
3-azidopropylamine, 3-azido-1-propanamine, 1-propanamine, 3-azido, 3-azido-propylamine, 1-azido-3-aminopropane, 1-amino-3-azidopropane, 3-aminopropyl azide, 3-azido-1-propylamine, 3-azidopropyl-1-amine
C(CN)CN=[N+]=[N-]
100.125
100.13
Liquid
88192-19-2
50°C
MFCD11046568
1993
OYBOVXXFJYJYPC-UHFFFAOYSA-N
3-azidopropan-1-amine
150110
≥95.0% (GC,T)

Chemical Identifiers

88192-19-2
100.125
OYBOVXXFJYJYPC-UHFFFAOYSA-N
150110
C(CN)CN=[N+]=[N-]
C3H8N4
MFCD11046568
3-azidopropylamine, 3-azido-1-propanamine, 1-propanamine, 3-azido, 3-azido-propylamine, 1-azido-3-aminopropane, 1-amino-3-azidopropane, 3-aminopropyl azide, 3-azido-1-propylamine, 3-azidopropyl-1-amine
3-azidopropan-1-amine
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions