Learn More
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, 98%, Thermo Scientific Chemicals
$112.85 - $334.19
Chemical Identifiers
CAS | 4568-71-2 |
---|---|
Molecular Formula | C13H16ClNOS |
Molecular Weight (g/mol) | 269.79 |
MDL Number | MFCD00011959 |
InChI Key | IWSVLBKHBJGMAA-UHFFFAOYSA-M |
Synonym | 3-benzyl-5-2-hydroxyethyl-4-methylthiazolium chloride, 3-benzyl-5-2-hydroxyethyl-4-methylthiazol-3-ium chloride, 3-benzyl-5-2-hydroxyethyl-4-methyl-1,3-thiazol-3-ium chloride, thiazolium, 5-2-hydroxyethyl-4-methyl-3-phenylmethyl-, chloride, 2-4-methyl-3-benzyl-1,3-thiazol-5-yl ethan-1-ol, chloride, acmc-1aqyn, 3-benzyl-5-2-hydroxyethyl thazolium chloride, 3-benzyl-4-methyl-5-hydroxyethylthiazolium chloride, 3-benzyl-5-hydroxyethyl-4-methylthiazolium chloride, 3-benzyl-5 2-hydroxyethyl-4-methylthiazolium chloride |
PubChem CID | 2833352 |
IUPAC Name | 2-(3-benzyl-4-methyl-1,3-thiazol-3-ium-5-yl)ethanol;chloride |
SMILES | [Cl-].CC1=C(CCO)SC=[N+]1CC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0875014
|
Thermo Scientific Chemicals
L0875014 |
25 g |
Each for $112.85
|
|
||||||
AAL0875022
|
Thermo Scientific Chemicals
L0875022 |
100 g |
Each for $334.19
|
|
||||||
Description
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, aliphatic aldehydes undergo the acyloin condensation in the presence of this catalyst and mild base. It acts as a catalyst for the addition of aliphatic and heterocyclic aldehydes to α,β-unsaturated ketones, nitriles and esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, aliphatic aldehydes undergo the acyloin condensation in the presence of this catalyst and mild base. It acts as a catalyst for the addition of aliphatic and heterocyclic aldehydes to α,β-unsaturated ketones, nitriles and esters.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with strong oxidizing agents.
Chemical Identifiers
4568-71-2 | |
269.79 | |
IWSVLBKHBJGMAA-UHFFFAOYSA-M | |
2833352 | |
[Cl-].CC1=C(CCO)SC=[N+]1CC1=CC=CC=C1 |
C13H16ClNOS | |
MFCD00011959 | |
3-benzyl-5-2-hydroxyethyl-4-methylthiazolium chloride, 3-benzyl-5-2-hydroxyethyl-4-methylthiazol-3-ium chloride, 3-benzyl-5-2-hydroxyethyl-4-methyl-1,3-thiazol-3-ium chloride, thiazolium, 5-2-hydroxyethyl-4-methyl-3-phenylmethyl-, chloride, 2-4-methyl-3-benzyl-1,3-thiazol-5-yl ethan-1-ol, chloride, acmc-1aqyn, 3-benzyl-5-2-hydroxyethyl thazolium chloride, 3-benzyl-4-methyl-5-hydroxyethylthiazolium chloride, 3-benzyl-5-hydroxyethyl-4-methylthiazolium chloride, 3-benzyl-5 2-hydroxyethyl-4-methylthiazolium chloride | |
2-(3-benzyl-4-methyl-1,3-thiazol-3-ium-5-yl)ethanol;chloride |
Specifications
4568-71-2 | |
C13H16ClNOS | |
25 g | |
Hygroscopic | |
Soluble in water. | |
[Cl-].CC1=C(CCO)SC=[N+]1CC1=CC=CC=C1 | |
269.79 | |
269.8 | |
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride |
142°C to 145°C | |
MFCD00011959 | |
3788921 | |
3-benzyl-5-2-hydroxyethyl-4-methylthiazolium chloride, 3-benzyl-5-2-hydroxyethyl-4-methylthiazol-3-ium chloride, 3-benzyl-5-2-hydroxyethyl-4-methyl-1,3-thiazol-3-ium chloride, thiazolium, 5-2-hydroxyethyl-4-methyl-3-phenylmethyl-, chloride, 2-4-methyl-3-benzyl-1,3-thiazol-5-yl ethan-1-ol, chloride, acmc-1aqyn, 3-benzyl-5-2-hydroxyethyl thazolium chloride, 3-benzyl-4-methyl-5-hydroxyethylthiazolium chloride, 3-benzyl-5-hydroxyethyl-4-methylthiazolium chloride, 3-benzyl-5 2-hydroxyethyl-4-methylthiazolium chloride | |
IWSVLBKHBJGMAA-UHFFFAOYSA-M | |
2-(3-benzyl-4-methyl-1,3-thiazol-3-ium-5-yl)ethanol;chloride | |
2833352 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 224-947-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only