missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Benzylamino)propylamine, 95%, Thermo Scientific™
$213.71 - $213.71
Chemical Identifiers
CAS | 13910-48-0 |
---|---|
Molecular Formula | C10H16N2 |
Molecular Weight (g/mol) | 164.25 |
MDL Number | MFCD00274361 |
InChI Key | RFLHDXQRFPJPRR-UHFFFAOYSA-N |
Synonym | n1-benzylpropane-1,3-diamine, 3-benzylamino propylamine, n-benzylpropane-1,3-diamine, 3-aminopropyl benzyl amine, n-benzyl-1,3-propanediamine, n-3-aminopropyl-n-benzylamine, 3-aminopropyl benzylamine, n-benzylpropan-1,3-diamin, n-benzyltrimethylenediamine, n-3-aminopropyl benzylamine |
PubChem CID | 83811 |
IUPAC Name | N'-benzylpropane-1,3-diamine |
SMILES | NCCCNCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6563503
|
Thermo Scientific Chemicals
H6563503 |
1 g |
Each for $213.71
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13910-48-0 | |
164.25 | |
RFLHDXQRFPJPRR-UHFFFAOYSA-N | |
83811 | |
NCCCNCC1=CC=CC=C1 |
C10H16N2 | |
MFCD00274361 | |
n1-benzylpropane-1,3-diamine, 3-benzylamino propylamine, n-benzylpropane-1,3-diamine, 3-aminopropyl benzyl amine, n-benzyl-1,3-propanediamine, n-3-aminopropyl-n-benzylamine, 3-aminopropyl benzylamine, n-benzylpropan-1,3-diamin, n-benzyltrimethylenediamine, n-3-aminopropyl benzylamine | |
N'-benzylpropane-1,3-diamine |
Specifications
13910-48-0 | |
C10H16N2 | |
MFCD00274361 | |
509316 | |
RFLHDXQRFPJPRR-UHFFFAOYSA-N | |
N'-benzylpropane-1,3-diamine | |
83811 | |
95% | |
3-(Benzylamino)propylamine |
Yellow | |
1.532 | |
1 g | |
n1-benzylpropane-1,3-diamine, 3-benzylamino propylamine, n-benzylpropane-1,3-diamine, 3-aminopropyl benzyl amine, n-benzyl-1,3-propanediamine, n-3-aminopropyl-n-benzylamine, 3-aminopropyl benzylamine, n-benzylpropan-1,3-diamin, n-benzyltrimethylenediamine, n-3-aminopropyl benzylamine | |
NCCCNCC1=CC=CC=C1 | |
164.25 | |
164.25 | |
Liquid |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 237-680-5
TSCA : No
Recommended Storage : Ambient temperatures