missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3'-Benzyloxyacetophenone 98.0+%, TCI America™
$131.49 - $449.83
Chemical Identifiers
CAS | 34068-01-4 |
---|---|
Molecular Formula | C15H14O2 |
Molecular Weight (g/mol) | 226.275 |
MDL Number | MFCD00026221 |
InChI Key | FGQMEAWGAUALJQ-UHFFFAOYSA-N |
Synonym | 1-3-benzyloxy phenyl ethanone, 3-benzyloxyacetophenone, 3'-benzyloxyacetophenone, 1-3-benzyloxy phenyl ethan-1-one, 1-3-phenylmethoxy phenyl ethan-1-one, 3'-benzyloxy acetophenone, ethanone, 1-3-phenylmethoxy phenyl, 1-acetyl-3-phenylmethoxy benzene |
PubChem CID | 98689 |
IUPAC Name | 1-(3-phenylmethoxyphenyl)ethanone |
SMILES | CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2 |
Chemical Identifiers
34068-01-4 | |
226.275 | |
FGQMEAWGAUALJQ-UHFFFAOYSA-N | |
98689 | |
CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2 |
C15H14O2 | |
MFCD00026221 | |
1-3-benzyloxy phenyl ethanone, 3-benzyloxyacetophenone, 3'-benzyloxyacetophenone, 1-3-benzyloxy phenyl ethan-1-one, 1-3-phenylmethoxy phenyl ethan-1-one, 3'-benzyloxy acetophenone, ethanone, 1-3-phenylmethoxy phenyl, 1-acetyl-3-phenylmethoxy benzene | |
1-(3-phenylmethoxyphenyl)ethanone |
Specifications
34068-01-4 | |
154°C | |
MFCD00026221 | |
1-3-benzyloxy phenyl ethanone, 3-benzyloxyacetophenone, 3'-benzyloxyacetophenone, 1-3-benzyloxy phenyl ethan-1-one, 1-3-phenylmethoxy phenyl ethan-1-one, 3'-benzyloxy acetophenone, ethanone, 1-3-phenylmethoxy phenyl, 1-acetyl-3-phenylmethoxy benzene | |
CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2 | |
226.275 | |
226.28 | |
Crystalline Lumps |
White-Yellow | |
C15H14O2 | |
5 g | |
FGQMEAWGAUALJQ-UHFFFAOYSA-N | |
1-(3-phenylmethoxyphenyl)ethanone | |
98689 | |
≥98.0% (GC) | |
3′-Benzyloxyacetophenone |
Safety and Handling
TSCA : No