missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Benzyloxycarbonylamino)azetidine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5283103

 View more versions of this product

Catalog No. AAH5283103


Explore available promotions

Description

Description

3-(Benzyloxycarbonylamino)azetidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Benzyloxycarbonylamino)azetidine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Specifications

3-(Benzyloxycarbonylamino)azetidine
C11H14N2O2
1 g
Slightly soluble in water.
C1C(CN1)NC(=O)OCC2=CC=CC=C2
206.245
206.24
914348-04-2
MFCD07368913
benzyl azetidin-3-ylcarbamate, 3-cbz-amino azetidine, 3-cbz-amino-azetidine, benzyl n-azetidin-3-yl carbamate, carbamic acid, n-3-azetidinyl-, phenylmethyl ester, azetidin-3-yl-carbamic acid benzyl ester, 3-benzyloxycarbonylamino azetidine, pubchem10159, 3-n-cbz-amino azetidine, benzyl azetidine-3-ylcarbamate
UWOOBFRZDNDUQB-UHFFFAOYSA-N
benzyl N-(azetidin-3-yl)carbamate
17750122
97%

Chemical Identifiers

914348-04-2
206.245
UWOOBFRZDNDUQB-UHFFFAOYSA-N
17750122
C1C(CN1)NC(=O)OCC2=CC=CC=C2
C11H14N2O2
MFCD07368913
benzyl azetidin-3-ylcarbamate, 3-cbz-amino azetidine, 3-cbz-amino-azetidine, benzyl n-azetidin-3-yl carbamate, carbamic acid, n-3-azetidinyl-, phenylmethyl ester, azetidin-3-yl-carbamic acid benzyl ester, 3-benzyloxycarbonylamino azetidine, pubchem10159, 3-n-cbz-amino azetidine, benzyl azetidine-3-ylcarbamate
benzyl N-(azetidin-3-yl)carbamate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only