Learn More
3-(Benzyloxycarbonylamino)azetidine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5283106
Description
3-(Benzyloxycarbonylamino)azetidine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(Benzyloxycarbonylamino)azetidine is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
3-(Benzyloxycarbonylamino)azetidine | |
C11H14N2O2 | |
5 g | |
Slightly soluble in water. | |
C1C(CN1)NC(=O)OCC2=CC=CC=C2 | |
206.245 | |
206.24 |
914348-04-2 | |
MFCD07368913 | |
benzyl azetidin-3-ylcarbamate, 3-cbz-amino azetidine, 3-cbz-amino-azetidine, benzyl n-azetidin-3-yl carbamate, carbamic acid, n-3-azetidinyl-, phenylmethyl ester, azetidin-3-yl-carbamic acid benzyl ester, 3-benzyloxycarbonylamino azetidine, pubchem10159, 3-n-cbz-amino azetidine, benzyl azetidine-3-ylcarbamate | |
UWOOBFRZDNDUQB-UHFFFAOYSA-N | |
benzyl N-(azetidin-3-yl)carbamate | |
17750122 | |
97% |
Chemical Identifiers
914348-04-2 | |
206.245 | |
UWOOBFRZDNDUQB-UHFFFAOYSA-N | |
17750122 | |
C1C(CN1)NC(=O)OCC2=CC=CC=C2 |
C11H14N2O2 | |
MFCD07368913 | |
benzyl azetidin-3-ylcarbamate, 3-cbz-amino azetidine, 3-cbz-amino-azetidine, benzyl n-azetidin-3-yl carbamate, carbamic acid, n-3-azetidinyl-, phenylmethyl ester, azetidin-3-yl-carbamic acid benzyl ester, 3-benzyloxycarbonylamino azetidine, pubchem10159, 3-n-cbz-amino azetidine, benzyl azetidine-3-ylcarbamate | |
benzyl N-(azetidin-3-yl)carbamate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only