missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Benzyloxymethyl)cyclobutanone, 97%, Thermo Scientific Chemicals

$430.97 - $430.97

Chemical Identifiers

CAS 172324-67-3
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD09055138
InChI Key FHBBBGYOVFMVIB-UHFFFAOYSA-N
PubChem CID 11819850
IUPAC Name 3-(phenylmethoxymethyl)cyclobutan-1-one
SMILES O=C1CC(COCC2=CC=CC=C2)C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6215903
View Documents
Thermo Scientific Chemicals
H6215903
1 g
Each for $430.97
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

172324-67-3
190.24
FHBBBGYOVFMVIB-UHFFFAOYSA-N
3-(phenylmethoxymethyl)cyclobutan-1-one
C12H14O2
MFCD09055138
11819850
O=C1CC(COCC2=CC=CC=C2)C1

Specifications

172324-67-3
MFCD09055138
3-benzyloxy methyl cyclobutanone, 3-benzyloxymethyl cyclobutanone, 3-benzyloxy methyl cyclobutan-1-one, cyclobutanone, 3-phenylmethoxy methyl, 3-benzyloxymethyl-cyclobutanone, 3-benzyloxymethyl cyclobutan-1-one, cyclobutanone,3-phenylmethoxy methyl
O=C1CC(COCC2=CC=CC=C2)C1
190.24
190.23
3-(Benzyloxymethyl)cyclobutanone
C12H14O2
1 g
FHBBBGYOVFMVIB-UHFFFAOYSA-N
3-(phenylmethoxymethyl)cyclobutan-1-one
11819850
97%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only