missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Boc-amino)benzeneboronic acid, 95%, Thermo Scientific Chemicals
$155.40 - $155.40
Chemical Identifiers
CAS | 380430-68-2 |
---|---|
Molecular Formula | C11H16BNO4 |
Molecular Weight (g/mol) | 237.062 |
MDL Number | MFCD03411945 |
InChI Key | CWLNHPXWZRALFS-UHFFFAOYSA-N |
Synonym | 3-boc-aminophenyl boronic acid, 3-n-boc-amino phenylboronic acid, 3-tert-butoxycarbonylamino phenylboronic acid, n-boc-3-aminophenylboronic acid, 3-tert-butoxycarbonyl amino phenylboronic acid, 3-boronoaniline, n-boc protected, 3-tert-butoxycarbonyl amino benzeneboronic acid, 3-boc-amino benzeneboronic acid, 3-tert-butoxycarbonylamino phenyl-boronic acid, 3-tert-butoxycarbonyl amino phenyl boronic acid |
PubChem CID | 2773228 |
IUPAC Name | [3-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid |
SMILES | B(C1=CC(=CC=C1)NC(=O)OC(C)(C)C)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2725803
|
Thermo Scientific Chemicals
H2725803 |
1 g |
Each for $155.40
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
380430-68-2 | |
237.062 | |
CWLNHPXWZRALFS-UHFFFAOYSA-N | |
2773228 | |
B(C1=CC(=CC=C1)NC(=O)OC(C)(C)C)(O)O |
C11H16BNO4 | |
MFCD03411945 | |
3-boc-aminophenyl boronic acid, 3-n-boc-amino phenylboronic acid, 3-tert-butoxycarbonylamino phenylboronic acid, n-boc-3-aminophenylboronic acid, 3-tert-butoxycarbonyl amino phenylboronic acid, 3-boronoaniline, n-boc protected, 3-tert-butoxycarbonyl amino benzeneboronic acid, 3-boc-amino benzeneboronic acid, 3-tert-butoxycarbonylamino phenyl-boronic acid, 3-tert-butoxycarbonyl amino phenyl boronic acid | |
[3-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid |
Specifications
380430-68-2 | |
C11H16BNO4 | |
1 g | |
CWLNHPXWZRALFS-UHFFFAOYSA-N | |
[3-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid | |
2773228 | |
95% |
∽155°C (decomposition) | |
MFCD03411945 | |
3-boc-aminophenyl boronic acid, 3-n-boc-amino phenylboronic acid, 3-tert-butoxycarbonylamino phenylboronic acid, n-boc-3-aminophenylboronic acid, 3-tert-butoxycarbonyl amino phenylboronic acid, 3-boronoaniline, n-boc protected, 3-tert-butoxycarbonyl amino benzeneboronic acid, 3-boc-amino benzeneboronic acid, 3-tert-butoxycarbonylamino phenyl-boronic acid, 3-tert-butoxycarbonyl amino phenyl boronic acid | |
B(C1=CC(=CC=C1)NC(=O)OC(C)(C)C)(O)O | |
237.062 | |
237.06 | |
3-(Boc-amino)benzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only