missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Boc-amino)benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
$167.84 - $514.99
Chemical Identifiers
CAS | 330793-09-4 |
---|---|
Molecular Formula | C17H26BNO4 |
Molecular Weight (g/mol) | 319.208 |
MDL Number | MFCD03789256 |
InChI Key | ANQAOGOIWVMGCH-UHFFFAOYSA-N |
Synonym | tert-butyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl-n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 3-n-boc-amino phenylboronic acid pinacol ester, 3-boc-amino benzeneboronic acid pinacol ester, tert-butyl n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 3-boc-aminophenylboronic acid, pinacol ester, tert-butyl n-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 3-tert-butoxycarbonylamino phenylboronic acid pinacol ester, 3-tert-butoxycarbonylamino phenylboronic acid, pinacol ester, 3-tert-butoxycarbonyl amino benzeneboronic acid, pinacol ester |
PubChem CID | 2773617 |
IUPAC Name | tert-butyl N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)NC(=O)OC(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2702603
|
Thermo Scientific Chemicals
H2702603 |
1 g |
Each for $167.84
|
|
|||||
AAH2702606
|
Thermo Scientific Chemicals
H2702606 |
5 g |
Each for $514.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
330793-09-4 | |
319.208 | |
ANQAOGOIWVMGCH-UHFFFAOYSA-N | |
2773617 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)NC(=O)OC(C)(C)C |
C17H26BNO4 | |
MFCD03789256 | |
tert-butyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl-n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 3-n-boc-amino phenylboronic acid pinacol ester, 3-boc-amino benzeneboronic acid pinacol ester, tert-butyl n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 3-boc-aminophenylboronic acid, pinacol ester, tert-butyl n-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 3-tert-butoxycarbonylamino phenylboronic acid pinacol ester, 3-tert-butoxycarbonylamino phenylboronic acid, pinacol ester, 3-tert-butoxycarbonyl amino benzeneboronic acid, pinacol ester | |
tert-butyl N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate |
Specifications
330793-09-4 | |
C17H26BNO4 | |
1 g | |
ANQAOGOIWVMGCH-UHFFFAOYSA-N | |
tert-butyl N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate | |
2773617 | |
97% |
118°C to 120°C | |
MFCD03789256 | |
tert-butyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl-n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 3-n-boc-amino phenylboronic acid pinacol ester, 3-boc-amino benzeneboronic acid pinacol ester, tert-butyl n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 3-boc-aminophenylboronic acid, pinacol ester, tert-butyl n-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 3-tert-butoxycarbonylamino phenylboronic acid pinacol ester, 3-tert-butoxycarbonylamino phenylboronic acid, pinacol ester, 3-tert-butoxycarbonyl amino benzeneboronic acid, pinacol ester | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)NC(=O)OC(C)(C)C | |
319.208 | |
319.21 | |
3-(Boc-amino)benzeneboronic acid pinacol ester |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only