missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-1,2-propanediol, 97%, Thermo Scientific Chemicals

$222.88 - $222.88

Chemical Identifiers

CAS 4704-77-2
Molecular Formula C3H7BrO2
Molecular Weight (g/mol) 154.99
MDL Number MFCD00004711
InChI Key SIBFQOUHOCRXDL-UHFFFAOYSA-N
PubChem CID 20827
ChEBI CHEBI:18719
IUPAC Name 3-bromopropane-1,2-diol
SMILES C(C(CBr)O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC209850050
SDS View Documents
Thermo Scientific Chemicals
209850050
5 mL Glass bottle
Each for $222.88
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4704-77-2
154.99
SIBFQOUHOCRXDL-UHFFFAOYSA-N
CHEBI:18719
C(C(CBr)O)O
C3H7BrO2
MFCD00004711
20827
3-bromopropane-1,2-diol

Specifications

4704-77-2
100.0
1.7700g/mL
>110°C
96% min. (GC)
C3H7BrO2
BrCH2CH(OH)CH2OH
5 mL
1.77
SIBFQOUHOCRXDL-UHFFFAOYSA-N
3-bromopropane-1,2-diol
20827
154.99
Viscous Liquid
96.0
Colorless to Yellow
72.0°C to 75.0°C (0.2 mmHg)
Authentic
Glass bottle
1.5165 to 1.5195
MFCD00004711
01, I, 248
3-bromo-1,2-propanediol, monobromoglycerol, alpha-bromohydrin, 1,2-propanediol, 3-bromo, bromodeoxyglycerol, glycerol bromohydrin, 1-bromo-2,3-propanediol, 3-bromodeoxyglycerol, rs-3-bromo-1,2-propanediol, 1,2-propanediol,3-bromo
C(C(CBr)O)O
154.99
CHEBI:18719
97%
3-Bromo-1, 2-propanediol
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 225-186-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only