missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-1,8-naphthalic anhydride, 97%, Thermo Scientific Chemicals

$200.88 - $200.88

Chemical Identifiers

CAS 24050-49-5
Molecular Formula C12H5BrO3
Molecular Weight (g/mol) 277.07
MDL Number MFCD00228162
InChI Key LYXFXCSFCWZGNZ-UHFFFAOYSA-N
Synonym 5-bromobenzo de isochromene-1,3-dione, 3-bromo-1,8-naphthalic anhydride, 5-bromo-1h,3h-benzo de isochromene-1,3-dione, pubchem24244, 5-bromo-benzo de isochromene-1,3-dione, 1h,3h-naphtho 1,8-cd pyran-1,3-dione,5-bromo, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 5-bromo, 11-bromo-3-oxatricyclo 7.3.1.0, 1 3 trideca-1 13 ,5,7,9,11-pentaene-2,4-dione, 7-bromo-3-oxatricyclo 7.3.1.0?,__ trideca-1 13 ,5,7,9,11-pentaene-2,4-dione, 11-bromo-3-oxatricyclo 7.3.1.0?, 1 3 trideca-1 13 ,5,7,9,11-pentaene-2,4-dione
PubChem CID 3907491
IUPAC Name 7-bromo-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
SMILES BrC1=CC2=CC=CC3=C2C(=C1)C(=O)OC3=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1768303
View Documents
Thermo Scientific Chemicals
L1768303
1 g
Each for $200.88
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

24050-49-5
277.07
LYXFXCSFCWZGNZ-UHFFFAOYSA-N
3907491
BrC1=CC2=CC=CC3=C2C(=C1)C(=O)OC3=O
C12H5BrO3
MFCD00228162
5-bromobenzo de isochromene-1,3-dione, 3-bromo-1,8-naphthalic anhydride, 5-bromo-1h,3h-benzo de isochromene-1,3-dione, pubchem24244, 5-bromo-benzo de isochromene-1,3-dione, 1h,3h-naphtho 1,8-cd pyran-1,3-dione,5-bromo, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 5-bromo, 11-bromo-3-oxatricyclo 7.3.1.0, 1 3 trideca-1 13 ,5,7,9,11-pentaene-2,4-dione, 7-bromo-3-oxatricyclo 7.3.1.0?,__ trideca-1 13 ,5,7,9,11-pentaene-2,4-dione, 11-bromo-3-oxatricyclo 7.3.1.0?, 1 3 trideca-1 13 ,5,7,9,11-pentaene-2,4-dione
7-bromo-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione

Specifications

24050-49-5
C12H5BrO3
1 g
Moisture sensitive
LYXFXCSFCWZGNZ-UHFFFAOYSA-N
7-bromo-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
3907491
97%
244°C to 246°C
MFCD00228162
180001
5-bromobenzo de isochromene-1,3-dione, 3-bromo-1,8-naphthalic anhydride, 5-bromo-1h,3h-benzo de isochromene-1,3-dione, pubchem24244, 5-bromo-benzo de isochromene-1,3-dione, 1h,3h-naphtho 1,8-cd pyran-1,3-dione,5-bromo, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 5-bromo, 11-bromo-3-oxatricyclo 7.3.1.0, 1 3 trideca-1 13 ,5,7,9,11-pentaene-2,4-dione, 7-bromo-3-oxatricyclo 7.3.1.0?,__ trideca-1 13 ,5,7,9,11-pentaene-2,4-dione, 11-bromo-3-oxatricyclo 7.3.1.0?, 1 3 trideca-1 13 ,5,7,9,11-pentaene-2,4-dione
BrC1=CC2=CC=CC3=C2C(=C1)C(=O)OC3=O
277.07
277.08
3-Bromo-1,8-naphthalic anhydride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only