missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-1-propanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 349701000

Catalog No. AC349701000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Bromo-1-propanol
627-18-9
100.0
62.0°C (5.0 mmHg)
Authentic
Glass bottle
1.4860 to 1.4900
MFCD00002942
01,356
3-bromo-1-propanol, trimethylene bromohydrin, 1-propanol, 3-bromo, 3-bromopropanol, 3-hydroxypropyl bromide, 1-bromo-3-propanol, 3-bromo-propan-1-ol, ccris 5979, 1-bromo-3-hydroxypropane, unii-3t4axh68fh
RQFUZUMFPRMVDX-UHFFFAOYSA-N
3-bromopropan-1-ol
12308
97%
97%
96.0
1.6000g/mL
65°C
96% min. (GC)
C3H7BrO
Br(CH2)3OH
100mL
1.6
Solubility in water: 167g/L water (20°C). Other solubilities: slightly soluble in ethanol and ether, soluble in most solvents
OCCCBr
138.99
138.99
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-986-6

RTECSNumber : UA7385000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only