missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1-(triisopropylsilyl)pyrrole 95.0+%, TCI America™

$123.03 - $1826.78

Chemical Identifiers

CAS 87630-36-2
Molecular Formula C13H24BrNSi
Molecular Weight (g/mol) 302.331
MDL Number MFCD01321136
InChI Key CNAUTMPRKBSDLH-UHFFFAOYSA-N
PubChem CID 2763302
IUPAC Name (3-bromopyrrol-1-yl)-tri(propan-2-yl)silane
SMILES CC(C)[Si](C(C)C)(C(C)C)N1C=CC(=C1)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B18991G
View Documents
TCI America
B18991G
1 g
Each for $123.03
Only null left
 
B18995G
View Documents
TCI America
B18995G
5 g
Each for $404.51
Only null left
 
B189925G
View Documents
TCI America
B189925G
25 g
Each for $1,826.78
Only null left
 
Description

Description

Specifications

Chemical Identifiers

87630-36-2
302.331
CNAUTMPRKBSDLH-UHFFFAOYSA-N
(3-bromopyrrol-1-yl)-tri(propan-2-yl)silane
C13H24BrNSi
MFCD01321136
2763302
CC(C)[Si](C(C)C)(C(C)C)N1C=CC(=C1)Br

Specifications

87630-36-2
C13H24BrNSi
1 g
CC(C)[Si](C(C)C)(C(C)C)N1C=CC(=C1)Br
302.331
302.33
Liquid
Yellow
MFCD01321136
CNAUTMPRKBSDLH-UHFFFAOYSA-N
(3-bromopyrrol-1-yl)-tri(propan-2-yl)silane
2763302
≥95.0% (GC)
3-Bromo-1-(triisopropylsilyl)pyrrole
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'