Learn More
3-Bromo-2-butanone, 95%, stabilized, Thermo Scientific Chemicals
$376.33 - $376.33
Chemical Identifiers
CAS | 814-75-5 |
---|---|
Molecular Formula | C4H7BrO |
Molecular Weight (g/mol) | 151 |
MDL Number | MFCD00013538 |
InChI Key | BNBOUFHCTIFWHN-UHFFFAOYSA-N |
Synonym | 3-bromo-2-butanone, 2-butanone, 3-bromo, 2-bromo-3-butanone, 3-bromo-butan-2-one, 1-bromoethyl methyl ketone, 2-bromobutan-2-one, 3-bromobutanone, 3-bromo-2-butanon, 2-bromo-3-oxobutane, 3-bromo-butan-2-on |
PubChem CID | 13142 |
IUPAC Name | 3-bromobutan-2-one |
SMILES | CC(C(=O)C)Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC402910050
|
Thermo Scientific Chemicals
402910050 |
5 g | Glass bottle |
Each for $376.33
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
814-75-5 | |
151 | |
BNBOUFHCTIFWHN-UHFFFAOYSA-N | |
13142 | |
CC(C(=O)C)Br |
C4H7BrO | |
MFCD00013538 | |
3-bromo-2-butanone, 2-butanone, 3-bromo, 2-bromo-3-butanone, 3-bromo-butan-2-one, 1-bromoethyl methyl ketone, 2-bromobutan-2-one, 3-bromobutanone, 3-bromo-2-butanon, 2-bromo-3-oxobutane, 3-bromo-butan-2-on | |
3-bromobutan-2-one |
Specifications
814-75-5 | |
100.0 | |
1.4380g/mL | |
51°C | |
94% min. (GC) | |
C4H7BrO | |
CH3COCHBrCH3 | |
5 g | |
3-bromo-2-butanone, 2-butanone, 3-bromo, 2-bromo-3-butanone, 3-bromo-butan-2-one, 1-bromoethyl methyl ketone, 2-bromobutan-2-one, 3-bromobutanone, 3-bromo-2-butanon, 2-bromo-3-oxobutane, 3-bromo-butan-2-on | |
CC(C(=O)C)Br | |
151 | |
13142 | |
95% | |
3-Bromo-2-butanone, 95% |
94.0 | |
Yellow to Brown | |
36.0°C (11.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4580 to 1.4600 | |
MFCD00013538 | |
1.438 | |
BNBOUFHCTIFWHN-UHFFFAOYSA-N | |
3-bromobutan-2-one | |
1% max. magnesium oxide | |
151 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to f
GHS Signal Word: Danger
EINECSNumber : 212-404-6
RTECSNumber : EL7005000
TSCA : TSCA
RUO – Research Use Only