missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-3-buten-1-ol, 97+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 368210500

 View more versions of this product

Catalog No. AC368210500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Bromo-3-buten-1-ol
97.0
1.5220g/mL
>110°C
97% min. (GC)
C4H7BrO
CH2=C(Br)CH2CH2OH
50g
1.522
RTKMFQOHBDVEBC-UHFFFAOYSA-N
3-bromobut-3-en-1-ol
533975
97+%
76334-36-6
100.0
175.0°C
Authentic
Glass bottle
1.4949
MFCD00154041
01, IV, 2100
3-bromo-3-buten-1-ol, 3-buten-1-ol, 3-bromo, 3bb, zlchem 79, acmc-20an90, 3-bromanylbut-3-en-1-ol
C=C(CCO)Br
151
151

Chemical Identifiers

76334-36-6
151
RTKMFQOHBDVEBC-UHFFFAOYSA-N
533975
C=C(CCO)Br
C4H7BrO
MFCD00154041
3-bromo-3-buten-1-ol, 3-buten-1-ol, 3-bromo, 3bb, zlchem 79, acmc-20an90, 3-bromanylbut-3-en-1-ol
3-bromobut-3-en-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only