missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Bromo-3-methyl-2-butanone 96.0+%, TCI America™
$185.65 - $549.05
Chemical Identifiers
CAS | 2648-71-7 |
---|---|
Molecular Formula | C5H9BrO |
Molecular Weight (g/mol) | 165.03 |
MDL Number | MFCD06661470 |
InChI Key | XHCWDDGZFBAXRT-UHFFFAOYSA-N |
Synonym | 1-Bromo-1-methylethyl Methyl Ketone |
PubChem CID | 344911 |
IUPAC Name | 3-bromo-3-methylbutan-2-one |
SMILES | CC(=O)C(C)(C)Br |
Chemical Identifiers
2648-71-7 | |
165.03 | |
XHCWDDGZFBAXRT-UHFFFAOYSA-N | |
344911 | |
CC(=O)C(C)(C)Br |
C5H9BrO | |
MFCD06661470 | |
1-Bromo-1-methylethyl Methyl Ketone | |
3-bromo-3-methylbutan-2-one |
Specifications
2648-71-7 | |
84°C | |
MFCD06661470 | |
1224 | |
XHCWDDGZFBAXRT-UHFFFAOYSA-N | |
3-bromo-3-methylbutan-2-one | |
344911 | |
≥96.0% (GC) | |
3-Bromo-3-methyl-2-butanone |
Yellow | |
C5H9BrO | |
5 g | |
1-Bromo-1-methylethyl Methyl Ketone | |
CC(=O)C(C)(C)Br | |
165.03 | |
165.03 | |
Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Freezer