missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Bromo-4-fluorophenylacetic acid, 96%, Thermo Scientific™
$40.64 - $82.47
Chemical Identifiers
CAS | 194019-11-9 |
---|---|
Molecular Formula | C8H6BrFO2 |
Molecular Weight (g/mol) | 233.04 |
MDL Number | MFCD04038373 |
InChI Key | XXFGIJYSXNXNAU-UHFFFAOYSA-N |
Synonym | 3-bromo-4-fluorophenylacetic acid, 2-3-bromo-4-fluorophenyl acetic acid, 3-bromo-4-fluorophenylaceticacid, 3-bromo-4-fluorophenyl acetic acid, benzeneacetic acid, 3-bromo-4-fluoro, 3-bromo-4-fluoro-phenyl-acetic acid, 2-3-bromo-4-fluoro-phenyl acetic acid, acmc-209exn, ksc497i4b, 61150-59-2 3-bromo-4-fluorophenylacetic acid |
PubChem CID | 3416339 |
IUPAC Name | 2-(3-bromo-4-fluorophenyl)acetic acid |
SMILES | OC(=O)CC1=CC(Br)=C(F)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6362203
|
Thermo Scientific Chemicals
H6362203 |
1 g |
N/A
|
|
|||||
AAH6362206
|
Thermo Scientific Chemicals
H6362206 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3-Bromo-4-fluorophenylacetic Acid is used as a reagent in the preparation of 4-oxo-1,4-dihydripyridine-3-carboxamide derivatives as inhibitors of Axl tyrosine kinase and anticancer agents.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
194019-11-9 | |
233.04 | |
XXFGIJYSXNXNAU-UHFFFAOYSA-N | |
3416339 | |
OC(=O)CC1=CC(Br)=C(F)C=C1 |
C8H6BrFO2 | |
MFCD04038373 | |
3-bromo-4-fluorophenylacetic acid, 2-3-bromo-4-fluorophenyl acetic acid, 3-bromo-4-fluorophenylaceticacid, 3-bromo-4-fluorophenyl acetic acid, benzeneacetic acid, 3-bromo-4-fluoro, 3-bromo-4-fluoro-phenyl-acetic acid, 2-3-bromo-4-fluoro-phenyl acetic acid, acmc-209exn, ksc497i4b, 61150-59-2 3-bromo-4-fluorophenylacetic acid | |
2-(3-bromo-4-fluorophenyl)acetic acid |
Specifications
194019-11-9 | |
C8H6BrFO2 | |
1 g | |
3-bromo-4-fluorophenylacetic acid, 2-3-bromo-4-fluorophenyl acetic acid, 3-bromo-4-fluorophenylaceticacid, 3-bromo-4-fluorophenyl acetic acid, benzeneacetic acid, 3-bromo-4-fluoro, 3-bromo-4-fluoro-phenyl-acetic acid, 2-3-bromo-4-fluoro-phenyl acetic acid, acmc-209exn, ksc497i4b, 61150-59-2 3-bromo-4-fluorophenylacetic acid | |
XXFGIJYSXNXNAU-UHFFFAOYSA-N | |
2-(3-bromo-4-fluorophenyl)acetic acid | |
3416339 | |
96% |
65°C to 69°C (decomposition) | |
MFCD04038373 | |
2446892 | |
Slightly soluble in water. | |
OC(=O)CC1=CC(Br)=C(F)C=C1 | |
233.04 | |
233.04 | |
3-Bromo-4-fluorophenylacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures