missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-4-heptanone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3209409

 View more versions of this product

Catalog No. AAH3209409


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Bromo-4-heptanone
1.248
75°C (167°F)
1.458
10 g
3-bromo-4-heptanone, 4-heptanone, 3-bromo
CCCC(=O)C(Br)CC
193.08
193.08
42330-10-9
39°C (1 mmHg)
C7H13BrO
MFCD09757526
UN3265
SFKVBRLKXVRUQW-UHFFFAOYNA-N
3-bromoheptan-4-one
11469593
98%

Chemical Identifiers

42330-10-9
193.08
SFKVBRLKXVRUQW-UHFFFAOYNA-N
11469593
CCCC(=O)C(Br)CC
C7H13BrO
MFCD09757526
3-bromo-4-heptanone, 4-heptanone, 3-bromo
3-bromoheptan-4-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only