missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

$62.26 - $681.16

Chemical Identifiers

CAS 34841-06-0
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00016599
InChI Key QMPNFQLVIGPNEI-UHFFFAOYSA-N
Synonym 3-bromo-p-anisaldehyde, benzaldehyde, 3-bromo-4-methoxy, 3-bromo-4-methoxy-benzaldehyde, 3-bromoanisaldehyde, 5-bromoanisaldehyde, 3-bromo-anisaldehyde, 5-bromo anisaldehyde, pubchem2643, acmc-209iat, 2-bromo-4-formylanisole
PubChem CID 98662
IUPAC Name 3-bromo-4-methoxybenzaldehyde
SMILES COC1=CC=C(C=O)C=C1Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1527406
View Documents
Thermo Scientific Chemicals
A1527406
5 g
Each for $62.26
 
AAA1527414
View Documents
Thermo Scientific Chemicals
A1527414
25 g
Each for $204.53
 
AAA1527422
View Documents
Thermo Scientific Chemicals
A1527422
100 g
Each for $681.16
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

34841-06-0
215.05
QMPNFQLVIGPNEI-UHFFFAOYSA-N
98662
COC1=CC=C(C=O)C=C1Br
C8H7BrO2
MFCD00016599
3-bromo-p-anisaldehyde, benzaldehyde, 3-bromo-4-methoxy, 3-bromo-4-methoxy-benzaldehyde, 3-bromoanisaldehyde, 5-bromoanisaldehyde, 3-bromo-anisaldehyde, 5-bromo anisaldehyde, pubchem2643, acmc-209iat, 2-bromo-4-formylanisole
3-bromo-4-methoxybenzaldehyde

Specifications

34841-06-0
108°C to 110°C (1 mmHg)
MFCD00016599
1636939
3-bromo-p-anisaldehyde, benzaldehyde, 3-bromo-4-methoxy, 3-bromo-4-methoxy-benzaldehyde, 3-bromoanisaldehyde, 5-bromoanisaldehyde, 3-bromo-anisaldehyde, 5-bromo anisaldehyde, pubchem2643, acmc-209iat, 2-bromo-4-formylanisole
COC1=CC=C(C=O)C=C1Br
215.05
215.05
3-Bromo-4-methoxybenzaldehyde
49°C to 53°C
C8H7BrO2
5 g
Air Sensitive
QMPNFQLVIGPNEI-UHFFFAOYSA-N
3-bromo-4-methoxybenzaldehyde
98662
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 252-241-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only