missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methoxypyridine, 97%

CAS: 82257-09-8 | C6H6BrNO | 188.024 g/mol

$75.38 - $875.59

Chemical Identifiers

CAS 82257-09-8
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD05664024
InChI Key DSFJKFHIMBVWAV-UHFFFAOYSA-N
PubChem CID 642827
IUPAC Name 3-bromo-4-methoxypyridine
SMILES COC1=C(C=NC=C1)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64296MD
View Documents
Thermo Scientific Chemicals
H64296MD
250 mg N/A
Only null left
 
AAH6429603
View Documents
Thermo Scientific Chemicals
H6429603
1 g N/A
Only null left
 
AAH6429606
View Documents
Thermo Scientific Chemicals
H6429606
5 g N/A
Only null left
 
Description

Description

3-Bromo-4-methoxypyridine acid is used as a intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-methoxypyridine acid is used as a intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water (9.8 g/L).

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

82257-09-8
188.024
DSFJKFHIMBVWAV-UHFFFAOYSA-N
3-bromo-4-methoxypyridine
C6H6BrNO
MFCD05664024
642827
COC1=C(C=NC=C1)Br

Specifications

82257-09-8
C6H6BrNO
250 mg
3-bromo-4-methoxy-pyridine, pyridine, 3-bromo-4-methoxy, pyridine,3-bromo-4-methoxy, pubchem6686, 4-methoxy-3-bromopyridine, acmc-209po0, abbypharma ap-17-5958
DSFJKFHIMBVWAV-UHFFFAOYSA-N
3-bromo-4-methoxypyridine
642827
97%
3-Bromo-4-methoxypyridine
Yellow
MFCD05664024
1423610
Slightly soluble in water (9.8g/L).
COC1=C(C=NC=C1)Br
188.024
188.02
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only