missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-4-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

$95.33 - $317.75

Chemical Identifiers

CAS 101420-81-9
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.02
MDL Number MFCD04117949
InChI Key KKPPNEJUUOQRLE-UHFFFAOYSA-N
Synonym 3-bromo-4-nitrobenzoic acid, 3-bromo-4-nitrobenzoicacid, benzoic acid,3-bromo-4-nitro, benzoic acid, 3-bromo-4-nitro, 3-bromo-4-nitrobenzoic acid 97, pubchem16301, acmc-209x3e, 4-nitro-3-bromobenzoic acid, 3-bromo-4-nitro-benzoic acid
PubChem CID 16218087
IUPAC Name 3-bromo-4-nitrobenzoic acid
SMILES OC(=O)C1=CC(Br)=C(C=C1)[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2726803
View Documents
Thermo Scientific Chemicals
H2726803
1 g
Each for $95.33
 
AAH2726806
View Documents
Thermo Scientific Chemicals
H2726806
5 g
Each for $317.75
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

101420-81-9
246.02
KKPPNEJUUOQRLE-UHFFFAOYSA-N
16218087
OC(=O)C1=CC(Br)=C(C=C1)[N+]([O-])=O
C7H4BrNO4
MFCD04117949
3-bromo-4-nitrobenzoic acid, 3-bromo-4-nitrobenzoicacid, benzoic acid,3-bromo-4-nitro, benzoic acid, 3-bromo-4-nitro, 3-bromo-4-nitrobenzoic acid 97, pubchem16301, acmc-209x3e, 4-nitro-3-bromobenzoic acid, 3-bromo-4-nitro-benzoic acid
3-bromo-4-nitrobenzoic acid

Specifications

101420-81-9
C7H4BrNO4
1 g
KKPPNEJUUOQRLE-UHFFFAOYSA-N
3-bromo-4-nitrobenzoic acid
16218087
97%
200°C to 204°C
MFCD04117949
3-bromo-4-nitrobenzoic acid, 3-bromo-4-nitrobenzoicacid, benzoic acid,3-bromo-4-nitro, benzoic acid, 3-bromo-4-nitro, 3-bromo-4-nitrobenzoic acid 97, pubchem16301, acmc-209x3e, 4-nitro-3-bromobenzoic acid, 3-bromo-4-nitro-benzoic acid
OC(=O)C1=CC(Br)=C(C=C1)[N+]([O-])=O
246.02
246.02
3-Bromo-4-nitrobenzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only