missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-chloro-2-hydroxybenzaldehyde, 97%

CAS: 19652-32-5 | C7H4BrClO2 | 235.461 g/mol

$111.58 - $383.07

Chemical Identifiers

CAS 19652-32-5
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.461
MDL Number MFCD00051690
InChI Key KNOYZLVIXXBBIB-UHFFFAOYSA-N
Synonym 3-bromo-5-chlorosalicylaldehyde, 3-bromo-5-chlorosalicylic aldehyde, benzaldehyde, 3-bromo-5-chloro-2-hydroxy, 3-bromo-5-chloro salicyclic aldehyde, 3-bromo-5-chloro-2-hydroxy-benzaldehyde, 3-bromo-5-chloro-o-hydroxybenzaldehyde, pubchem3591, acmc-1bs9s, akos bb-6948, knoyzlvixxbbib-uhfffaoysa
PubChem CID 519676
IUPAC Name 3-bromo-5-chloro-2-hydroxybenzaldehyde
SMILES C1=C(C=C(C(=C1Br)O)C=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1344906
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
A1344906
5 g
Each for $111.58
Only null left
 
AAA1344914
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
A1344914
25 g
Each for $383.07
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

19652-32-5
235.461
KNOYZLVIXXBBIB-UHFFFAOYSA-N
519676
C1=C(C=C(C(=C1Br)O)C=O)Cl
C7H4BrClO2
MFCD00051690
3-bromo-5-chlorosalicylaldehyde, 3-bromo-5-chlorosalicylic aldehyde, benzaldehyde, 3-bromo-5-chloro-2-hydroxy, 3-bromo-5-chloro salicyclic aldehyde, 3-bromo-5-chloro-2-hydroxy-benzaldehyde, 3-bromo-5-chloro-o-hydroxybenzaldehyde, pubchem3591, acmc-1bs9s, akos bb-6948, knoyzlvixxbbib-uhfffaoysa
3-bromo-5-chloro-2-hydroxybenzaldehyde

Specifications

19652-32-5
C7H4BrClO2
5 g
Air Sensitive
KNOYZLVIXXBBIB-UHFFFAOYSA-N
3-bromo-5-chloro-2-hydroxybenzaldehyde
519676
97%
84°C to 88°C
MFCD00051690
2362159
3-bromo-5-chlorosalicylaldehyde, 3-bromo-5-chlorosalicylic aldehyde, benzaldehyde, 3-bromo-5-chloro-2-hydroxy, 3-bromo-5-chloro salicyclic aldehyde, 3-bromo-5-chloro-2-hydroxy-benzaldehyde, 3-bromo-5-chloro-o-hydroxybenzaldehyde, pubchem3591, acmc-1bs9s, akos bb-6948, knoyzlvixxbbib-uhfffaoysa
C1=C(C=C(C(=C1Br)O)C=O)Cl
235.461
235.47
3-Bromo-5-chloro-2-hydroxybenzaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only