missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-5-methoxypyridine 97.0+%, TCI America™
$65.24 - $259.11
Chemical Identifiers
| CAS | 50720-12-2 |
|---|---|
| Molecular Formula | C6H6BrNO |
| Molecular Weight (g/mol) | 188.024 |
| MDL Number | MFCD00234169 |
| InChI Key | FZWUIWQMJFAWJW-UHFFFAOYSA-N |
| Synonym | 3-bromo-5-methoxy-pyridine, 5-methoxy-3-bromopyridine, 5-bromo-3-methoxypyridine, pyridine, 3-bromo-5-methoxy, 5-bromopyridin-3-yl methyl ether, 3-methoxy-5-bromopyridine, 3-bromo-5-methoxy pyridine, 3-brom-5-methoxypyridin, zlchem 930, pubchem5085 |
| PubChem CID | 817163 |
| IUPAC Name | 3-bromo-5-methoxypyridine |
| SMILES | COC1=CC(=CN=C1)Br |
Chemical Identifiers
| 50720-12-2 | |
| 188.024 | |
| FZWUIWQMJFAWJW-UHFFFAOYSA-N | |
| 817163 | |
| COC1=CC(=CN=C1)Br |
| C6H6BrNO | |
| MFCD00234169 | |
| 3-bromo-5-methoxy-pyridine, 5-methoxy-3-bromopyridine, 5-bromo-3-methoxypyridine, pyridine, 3-bromo-5-methoxy, 5-bromopyridin-3-yl methyl ether, 3-methoxy-5-bromopyridine, 3-bromo-5-methoxy pyridine, 3-brom-5-methoxypyridin, zlchem 930, pubchem5085 | |
| 3-bromo-5-methoxypyridine |
Specifications
| 50720-12-2 | |
| C6H6BrNO | |
| 1 g | |
| FZWUIWQMJFAWJW-UHFFFAOYSA-N | |
| 3-bromo-5-methoxypyridine | |
| 817163 | |
| ≥97.0% (T) | |
| 3-Bromo-5-methoxypyridine |
| White-Yellow | |
| MFCD00234169 | |
| 3-bromo-5-methoxy-pyridine, 5-methoxy-3-bromopyridine, 5-bromo-3-methoxypyridine, pyridine, 3-bromo-5-methoxy, 5-bromopyridin-3-yl methyl ether, 3-methoxy-5-bromopyridine, 3-bromo-5-methoxy pyridine, 3-brom-5-methoxypyridin, zlchem 930, pubchem5085 | |
| COC1=CC(=CN=C1)Br | |
| 188.024 | |
| 188.02 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No