missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-5-methoxytoluene 98.0+%, TCI America™
$41.02 - $91.33
Chemical Identifiers
| CAS | 29578-83-4 |
|---|---|
| Molecular Formula | C8H9BrO |
| Molecular Weight (g/mol) | 201.063 |
| MDL Number | MFCD08061916 |
| InChI Key | AOEVRCZZWJWKPG-UHFFFAOYSA-N |
| Synonym | 3-Bromo-5-methylanisole |
| PubChem CID | 10679554 |
| IUPAC Name | 1-bromo-3-methoxy-5-methylbenzene |
| SMILES | CC1=CC(=CC(=C1)Br)OC |
Chemical Identifiers
| 29578-83-4 | |
| 201.063 | |
| AOEVRCZZWJWKPG-UHFFFAOYSA-N | |
| 10679554 | |
| CC1=CC(=CC(=C1)Br)OC |
| C8H9BrO | |
| MFCD08061916 | |
| 3-Bromo-5-methylanisole | |
| 1-bromo-3-methoxy-5-methylbenzene |
Specifications
| 29578-83-4 | |
| C8H9BrO | |
| 1 g | |
| AOEVRCZZWJWKPG-UHFFFAOYSA-N | |
| 1-bromo-3-methoxy-5-methylbenzene | |
| 10679554 | |
| ≥98.0% (GC) | |
| 3-Bromo-5-methoxytoluene |
| 122°C | |
| MFCD08061916 | |
| 3-Bromo-5-methylanisole | |
| CC1=CC(=CC(=C1)Br)OC | |
| 201.063 | |
| 201.06 | |
| Liquid |
Safety and Handling
TSCA : No