missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Bromo-5-nitrosalicylaldehyde 95.0+%, TCI America™
$91.70 - $335.63
Chemical Identifiers
CAS | 16789-84-7 |
---|---|
Molecular Formula | C7H4BrNO4 |
Molecular Weight (g/mol) | 246.02 |
MDL Number | MFCD00051833 |
InChI Key | BESBCGANGAEHPM-UHFFFAOYSA-N |
Synonym | 3-bromo-2-hydroxy-5-nitrobenzaldehyde, 3-bromo-5-nitrosalicylaldehyde, 3-bromo-2-hydroxy-5-nitrobenzenecarbaldehyde, 2-hydroxy-3-bromo-5-nitrobenzaldehyde, 3-bromo-2-hydroxy-5-nitro-benzaldehyde, acmc-1cfaz, 3-bromo-5-nitro-salicylaldehyde, benzaldehyde,3-bromo-2-hydroxy-5-nitro, 3-bromanyl-5-nitro-2-oxidanyl-benzaldehyde |
PubChem CID | 519307 |
IUPAC Name | 3-bromo-2-hydroxy-5-nitrobenzaldehyde |
SMILES | OC1=C(Br)C=C(C=C1C=O)[N+]([O-])=O |
Chemical Identifiers
16789-84-7 | |
246.02 | |
BESBCGANGAEHPM-UHFFFAOYSA-N | |
519307 | |
OC1=C(Br)C=C(C=C1C=O)[N+]([O-])=O |
C7H4BrNO4 | |
MFCD00051833 | |
3-bromo-2-hydroxy-5-nitrobenzaldehyde, 3-bromo-5-nitrosalicylaldehyde, 3-bromo-2-hydroxy-5-nitrobenzenecarbaldehyde, 2-hydroxy-3-bromo-5-nitrobenzaldehyde, 3-bromo-2-hydroxy-5-nitro-benzaldehyde, acmc-1cfaz, 3-bromo-5-nitro-salicylaldehyde, benzaldehyde,3-bromo-2-hydroxy-5-nitro, 3-bromanyl-5-nitro-2-oxidanyl-benzaldehyde | |
3-bromo-2-hydroxy-5-nitrobenzaldehyde |
Specifications
16789-84-7 | |
Yellow | |
MFCD00051833 | |
2811 | |
BESBCGANGAEHPM-UHFFFAOYSA-N | |
3-bromo-2-hydroxy-5-nitrobenzaldehyde | |
519307 | |
≥95.0% (GC,T) | |
3-Bromo-5-nitrosalicylaldehyde |
149°C | |
C7H4BrNO4 | |
1 g | |
3-bromo-2-hydroxy-5-nitrobenzaldehyde, 3-bromo-5-nitrosalicylaldehyde, 3-bromo-2-hydroxy-5-nitrobenzenecarbaldehyde, 2-hydroxy-3-bromo-5-nitrobenzaldehyde, 3-bromo-2-hydroxy-5-nitro-benzaldehyde, acmc-1cfaz, 3-bromo-5-nitro-salicylaldehyde, benzaldehyde,3-bromo-2-hydroxy-5-nitro, 3-bromanyl-5-nitro-2-oxidanyl-benzaldehyde | |
OC1=C(Br)C=C(C=C1C=O)[N+]([O-])=O | |
246.02 | |
246.02 | |
Crystalline Powder |
Safety and Handling
TSCA : No