missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-6-chloroimidazo[1,2-a]pyridine, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3555803

 View more versions of this product

Catalog No. AAH3555803


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Bromo-6-chloroimidazo[1,2-a]pyridine
105°C to 107°C
MFCD08667858
3-bromo-6-chloroimidazo 1,2-a pyridine, 6-chloro-3-bromo-imidazo 1,2-a pyridine, acmc-209qvk, 3-bromo-6-chloro-imidazo 1,2-a pyridine, imidazo 1,2-a pyridine, 3-bromo-6-chloro
C1=CC2=NC=C(N2C=C1Cl)Br
231.477
231.48
886371-28-4
C7H4BrClN2
1 g
LIBKDAUSMLPFBF-UHFFFAOYSA-N
3-bromo-6-chloroimidazo[1,2-a]pyridine
45790042
95%

Chemical Identifiers

886371-28-4
231.477
LIBKDAUSMLPFBF-UHFFFAOYSA-N
45790042
C1=CC2=NC=C(N2C=C1Cl)Br
C7H4BrClN2
MFCD08667858
3-bromo-6-chloroimidazo 1,2-a pyridine, 6-chloro-3-bromo-imidazo 1,2-a pyridine, acmc-209qvk, 3-bromo-6-chloro-imidazo 1,2-a pyridine, imidazo 1,2-a pyridine, 3-bromo-6-chloro
3-bromo-6-chloroimidazo[1,2-a]pyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only