Learn More
3-Bromo-N,N-dimethylaniline, 97%
CAS: 16518-62-0 | C8H10BrN | 200.08 g/mol
$167.34 - $566.66
Chemical Identifiers
| CAS | 16518-62-0 |
|---|---|
| Molecular Formula | C8H10BrN |
| Molecular Weight (g/mol) | 200.08 |
| MDL Number | MFCD00045020 |
| InChI Key | USEXQPWLCGBYNT-UHFFFAOYSA-N |
| Synonym | benzenamine, 3-bromo-n,n-dimethyl, 3-bromophenyl dimethylamine, n,n-dimethyl 3-bromoaniline, 3-bromo-n,n-dimethylbenzenamine, n,n-dimethyl-3-bromoaniline, m-bromodimethyl aniline, 3-bromo-dimethylaniline, m-bromodimethylaniline, pubchem3760, acmc-1c80w |
| PubChem CID | 140102 |
| IUPAC Name | 3-bromo-N,N-dimethylaniline |
| SMILES | CN(C)C1=CC=CC(Br)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1727906
|
Thermo Scientific Chemicals
L1727906 |
5 g |
Each for $167.34
|
|
|||||
|
AAL1727914
|
Thermo Scientific Chemicals
L1727914 |
25 g |
Each for $566.66
|
|
|||||
Description
3-Bromo-N,N-dimethylaniline is involved in fluorination using metal fluoride in the presence of 2-(di-1-adamantylphosphino)-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl (AdBrettPhos)-based Pd precatalyst, used to prepare 3-fluoro-N,N- dimethylaniline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Bromo-N,N-dimethylaniline is involved in fluorination using metal fluoride in the presence of 2-(di-1-adamantylphosphino)-3,6-dimethoxy-2′,4′,6′-tri-i-propyl-1,1′-biphenyl (AdBrettPhos)-based Pd precatalyst, used to prepare 3-fluoro-N,N- dimethylaniline.
Solubility
Miscible with chloroform, dichloromethane and methanol.
Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents and strong acids.
Chemical Identifiers
| 16518-62-0 | |
| 200.08 | |
| USEXQPWLCGBYNT-UHFFFAOYSA-N | |
| 140102 | |
| CN(C)C1=CC=CC(Br)=C1 |
| C8H10BrN | |
| MFCD00045020 | |
| benzenamine, 3-bromo-n,n-dimethyl, 3-bromophenyl dimethylamine, n,n-dimethyl 3-bromoaniline, 3-bromo-n,n-dimethylbenzenamine, n,n-dimethyl-3-bromoaniline, m-bromodimethyl aniline, 3-bromo-dimethylaniline, m-bromodimethylaniline, pubchem3760, acmc-1c80w | |
| 3-bromo-N,N-dimethylaniline |
Specifications
| 16518-62-0 | |
| 1.365 | |
| C8H10BrN | |
| MFCD00045020 | |
| 2079908 | |
| Miscible with chloroform,dichloromethane and methanol. | |
| CN(C)C1=CC=CC(Br)=C1 | |
| 200.08 | |
| 200.08 | |
| 3-Bromo-N,N-dimethylaniline |
| 11°C | |
| 126°C (14 mmHg) | |
| 1.6015 | |
| 5 g | |
| benzenamine, 3-bromo-n,n-dimethyl, 3-bromophenyl dimethylamine, n,n-dimethyl 3-bromoaniline, 3-bromo-n,n-dimethylbenzenamine, n,n-dimethyl-3-bromoaniline, m-bromodimethyl aniline, 3-bromo-dimethylaniline, m-bromodimethylaniline, pubchem3760, acmc-1c80w | |
| USEXQPWLCGBYNT-UHFFFAOYSA-N | |
| 3-bromo-N,N-dimethylaniline | |
| 140102 | |
| 97% |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only