missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromoanisole, 99+%, Thermo Scientific™

Chemical Identifiers

CAS 2398-37-0
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00000081
InChI Key PLDWAJLZAAHOGG-UHFFFAOYSA-N
Synonym 3-bromoanisole, m-bromoanisole, 3-methoxybromobenzene, 3-bromo anisole, m-bromomethoxybenzene, benzene, 1-bromo-3-methoxy, anisole, m-bromo, 3-methoxy-1-bromobenzene, m-methoxybromobenzene, meta-bromoanisole
PubChem CID 16971
IUPAC Name 1-bromo-3-methoxybenzene
SMILES COC1=CC=CC(Br)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC106620250
SDS View Documents
Thermo Scientific Chemicals
106620250
25mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC106621000
SDS View Documents
Thermo Scientific Chemicals
106621000
100mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2398-37-0
187.04
PLDWAJLZAAHOGG-UHFFFAOYSA-N
16971
COC1=CC=CC(Br)=C1
C7H7BrO
MFCD00000081
3-bromoanisole, m-bromoanisole, 3-methoxybromobenzene, 3-bromo anisole, m-bromomethoxybenzene, benzene, 1-bromo-3-methoxy, anisole, m-bromo, 3-methoxy-1-bromobenzene, m-methoxybromobenzene, meta-bromoanisole
1-bromo-3-methoxybenzene

Specifications

2398-37-0
100.0
210.0°C to 211.0°C
Authentic
Glass bottle
1.5630 to 1.5650
MFCD00000081
06, 198
3-bromoanisole, m-bromoanisole, 3-methoxybromobenzene, 3-bromo anisole, m-bromomethoxybenzene, benzene, 1-bromo-3-methoxy, anisole, m-bromo, 3-methoxy-1-bromobenzene, m-methoxybromobenzene, meta-bromoanisole
COC1=CC=CC(Br)=C1
187.04
187.04
3-Bromoanisole, 99+%
99.0
1.4700g/mL
93°C
99% min. (GC)
C7H7BrO
BrC6H4OCH3
25mL
1.47
PLDWAJLZAAHOGG-UHFFFAOYSA-N
1-bromo-3-methoxybenzene
16971
99+%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 219-264-5

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only