missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromobenzoyl chloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 397450050

 View more versions of this product

Catalog No. AC397450050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Bromobenzoyl chloride
96.0
1.6660g/mL
107°C
Glass bottle
BrC6H4COCl
5 g
m-bromobenzoyl chloride, benzoyl chloride, 3-bromo, 3-bromobenzoylchloride, benzoyl chloride, m-bromo, 3-bromobenzoic acid chloride, attercop-chm at106527, 3-brom-benzoylchlorid, 3-bromo-benzoylchlorid, 3-bromobenzoic chloride, pubchem13574
C1=CC(=CC(=C1)Br)C(=O)Cl
219.46
219.46
1711-09-7
100.0
247°C to 251°C
96% min. (GC)
C7H4BrClO
MFCD00000669
1.666
PBOOZQFGWNZNQE-UHFFFAOYSA-N
3-bromobenzoyl chloride
74377
97%

Chemical Identifiers

1711-09-7
219.46
PBOOZQFGWNZNQE-UHFFFAOYSA-N
74377
C1=CC(=CC(=C1)Br)C(=O)Cl
C7H4BrClO
MFCD00000669
m-bromobenzoyl chloride, benzoyl chloride, 3-bromo, 3-bromobenzoylchloride, benzoyl chloride, m-bromo, 3-bromobenzoic acid chloride, attercop-chm at106527, 3-brom-benzoylchlorid, 3-bromo-benzoylchlorid, 3-bromobenzoic chloride, pubchem13574
3-bromobenzoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 216-978-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only