Learn More
3-Bromobenzyl bromide, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 181780050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Bromobenzyl bromide | |
823-78-9 | |
100.0 | |
White | |
>110°C | |
94% min. (GC) | |
C7H6Br2 | |
MFCD00000176 | |
05, 308 | |
Solubility in water: insoluble. Other solubilities: soluble in methanol | |
C1=CC(=CC(=C1)Br)CBr | |
249.93 | |
249.93 | |
Adhering Crystals or Crystalline Powder |
95% | |
94.0 | |
37.0°C to 42.0°C | |
130.0°C (12.0 mmHg) | |
Authentic | |
Glass bottle | |
BrC6H4CH2Br | |
5 g | |
3-bromobenzyl bromide, 1-bromo-3-bromomethyl benzene, m-bromobenzyl bromide, 3-bromobenzylbromide, benzene, 1-bromo-3-bromomethyl, alpha,3-dibromotoluene, alpha,m-dibromotoluene, 3-bromo benzyl bromide, 1-bromo-3-bromomethyl-benzene, alpha-3-dibromotoluene | |
ZPCJPJQUVRIILS-UHFFFAOYSA-N | |
1-bromo-3-(bromomethyl)benzene | |
69979 | |
95% |
Chemical Identifiers
823-78-9 | |
249.93 | |
ZPCJPJQUVRIILS-UHFFFAOYSA-N | |
69979 | |
C1=CC(=CC(=C1)Br)CBr |
C7H6Br2 | |
MFCD00000176 | |
3-bromobenzyl bromide, 1-bromo-3-bromomethyl benzene, m-bromobenzyl bromide, 3-bromobenzylbromide, benzene, 1-bromo-3-bromomethyl, alpha,3-dibromotoluene, alpha,m-dibromotoluene, 3-bromo benzyl bromide, 1-bromo-3-bromomethyl-benzene, alpha-3-dibromotoluene | |
1-bromo-3-(bromomethyl)benzene |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 212-519-1
TSCA : TSCA
RUO – Research Use Only