missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromochromone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1459103

 View more versions of this product

Catalog No. AAA1459103


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Bromochromone
94°C to 98°C
MFCD00017337
125684
IQIGYNPOESZBDJ-UHFFFAOYSA-N
3-bromochromen-4-one
521256
97%
49619-82-1
C9H5BrO2
1 g
3-bromochromone, 3-bromo-4h-chromen-4-one, 3-bromo-4-oxo-4h-1-benzopyran, 3-bromo chromone, maybridge1_006104, acmc-209kh2, 3-bromo-4h-chromen-4-one #, 3-bromo-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,3-bromo
C1=CC=C2C(=C1)C(=O)C(=CO2)Br
225.041
225.05

Chemical Identifiers

49619-82-1
225.041
IQIGYNPOESZBDJ-UHFFFAOYSA-N
521256
C1=CC=C2C(=C1)C(=O)C(=CO2)Br
C9H5BrO2
MFCD00017337
3-bromochromone, 3-bromo-4h-chromen-4-one, 3-bromo-4-oxo-4h-1-benzopyran, 3-bromo chromone, maybridge1_006104, acmc-209kh2, 3-bromo-4h-chromen-4-one #, 3-bromo-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,3-bromo
3-bromochromen-4-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only