missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromocyclohexene, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1037022

 View more versions of this product

Catalog No. AAA1037022


Explore available promotions

Description

Description

In peptide synthesis, can be used to protect the hydroxyl group of serine and threonine as the cyclohexyl (Chx) ether, by reaction with the sodium alkoxide (generated with NaH), followed by hydrogenation. The group is stable to TFA and 20% piperidine in DMF, but can be cleaved with TfOH in TFA, in the presence of thioanisole as a cation scavenger. It is also used in the synthesis of N-tert-butoxycarbonyl-O-cyclohexyl-L-tyrosine.1 It was also used in the synthesis of enantiopure cyclohexitols such as muco-quercitol, D-chiro-inocitol and allo-inocitol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In peptide synthesis, can be used to protect the hydroxyl group of serine and threonine as the cyclohexyl (Chx) ether, by reaction with the sodium alkoxide (generated with NaH), followed by hydrogenation. The group is stable to TFA and 20% piperidine in DMF, but can be cleaved with TfOH in TFA, in the presence of thioanisole as a cation scavenger. It is also used in the synthesis of N-tert-butoxycarbonyl-O-cyclohexyl-L-tyrosine.1 It was also used in the synthesis of enantiopure cyclohexitols such as muco-quercitol, D-chiro-inocitol and allo-inocitol.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

3-Bromocyclohexene
1521-51-3
64°C to 65°C (15 mmHg)
C6H9Br
MFCD00013775
UN1993
3-bromocyclohex-1-ene, cyclohexene, 3-bromo, 1-bromo-2-cyclohexene, 3-bromo-1-cyclohexene, 1,2,3,4-tetrahydrobromobenzene, 3-bromo-cyclohexene, 2-cyclohexenyl bromide, 2-cyclohexen-1-yl bromide, 3-bromo cyclohexene, 3-bromanylcyclohexene
AJKDUJRRWLQXHM-UHFFFAOYSA-N
3-bromocyclohexene
137057
95%
Stabilized with 300 to 1000ppm Propylene oxide
1.4
54°C (129°F)
1.531
100 g
635953
Not miscible or difficult to mix in water.
C1CC=CC(C1)Br
161.042
161.05

Chemical Identifiers

1521-51-3
161.042
AJKDUJRRWLQXHM-UHFFFAOYSA-N
137057
C1CC=CC(C1)Br
C6H9Br
MFCD00013775
3-bromocyclohex-1-ene, cyclohexene, 3-bromo, 1-bromo-2-cyclohexene, 3-bromo-1-cyclohexene, 1,2,3,4-tetrahydrobromobenzene, 3-bromo-cyclohexene, 2-cyclohexenyl bromide, 2-cyclohexen-1-yl bromide, 3-bromo cyclohexene, 3-bromanylcyclohexene
3-bromocyclohexene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P374-P380-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only