missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromophenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

$180.42 - $1716.60

Chemical Identifiers

CAS 27246-81-7
Molecular Formula C6H8BrClN2
Molecular Weight (g/mol) 223.50
MDL Number MFCD00012933
InChI Key RPYIPFXHIKXRKS-UHFFFAOYSA-N
Synonym 3-bromophenylhydrazine hydrochloride, 3-bromophenyl hydrazine hydrochloride, 1-3-bromophenyl hydrazine hydrochloride, m-bromophenyl hydrazine hydrochloride, hydrazine, m-bromophenyl-, monohydrochloride, 3-bromophenylhydrazinehydrochloride, 3-bromophenyl hydrazine hcl, hydrazine, 3-bromophenyl-, monohydrochloride, pubchem23212, acmc-1clhx
PubChem CID 33757
IUPAC Name (3-bromophenyl)hydrazine;hydrochloride
SMILES [H+].[Cl-].NNC1=CC=CC(Br)=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2351006
View Documents
Thermo Scientific Chemicals
B2351006
5 g
Each for $180.42
 
AAB2351014
View Documents
Thermo Scientific Chemicals
B2351014
25 g
Each for $580.73
 
AAB2351022
View Documents
Thermo Scientific Chemicals
B2351022
100 g
Each for $1,716.60
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

27246-81-7
223.50
RPYIPFXHIKXRKS-UHFFFAOYSA-N
33757
[H+].[Cl-].NNC1=CC=CC(Br)=C1
C6H8BrClN2
MFCD00012933
3-bromophenylhydrazine hydrochloride, 3-bromophenyl hydrazine hydrochloride, 1-3-bromophenyl hydrazine hydrochloride, m-bromophenyl hydrazine hydrochloride, hydrazine, m-bromophenyl-, monohydrochloride, 3-bromophenylhydrazinehydrochloride, 3-bromophenyl hydrazine hcl, hydrazine, 3-bromophenyl-, monohydrochloride, pubchem23212, acmc-1clhx
(3-bromophenyl)hydrazine;hydrochloride

Specifications

27246-81-7
C6H8BrClN2
5 g
3-bromophenylhydrazine hydrochloride, 3-bromophenyl hydrazine hydrochloride, 1-3-bromophenyl hydrazine hydrochloride, m-bromophenyl hydrazine hydrochloride, hydrazine, m-bromophenyl-, monohydrochloride, 3-bromophenylhydrazinehydrochloride, 3-bromophenyl hydrazine hcl, hydrazine, 3-bromophenyl-, monohydrochloride, pubchem23212, acmc-1clhx
[H+].[Cl-].NNC1=CC=CC(Br)=C1
223.50
223.51
3-Bromophenylhydrazine hydrochloride
∼225°C (decomposition)
MFCD00012933
3565829
RPYIPFXHIKXRKS-UHFFFAOYSA-N
(3-bromophenyl)hydrazine;hydrochloride
33757
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 248-362-0

RTECSNumber : MV0815000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only