Learn More
3-Bromopropionitrile, 97%
CAS: 2417-90-5 | C3H4BrN | 133.976 g/mol
$83.85 - $218.90
Chemical Identifiers
| CAS | 2417-90-5 |
|---|---|
| Molecular Formula | C3H4BrN |
| Molecular Weight (g/mol) | 133.976 |
| MDL Number | MFCD00001951 |
| InChI Key | CQZIEDXCLQOOEH-UHFFFAOYSA-N |
| Synonym | 3-bromopropionitrile, propanenitrile, 3-bromo, 2-bromoethyl cyanide, 3-bromopropiononitrile, 1-bromo-2-cyanoethane, beta-bromopropionitrile, 2-cyanoethyl bromide, propionitrile, 3-bromo, usaf do-51, beta-bromopriopionitrile |
| PubChem CID | 17020 |
| IUPAC Name | 3-bromopropanenitrile |
| SMILES | C(CBr)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2399814
|
Thermo Scientific Chemicals
B2399814 |
25 g |
Each for $83.85
|
|
|||||
|
AAB2399822
|
Thermo Scientific Chemicals
B2399822 |
100 g |
Each for $218.90
|
|
|||||
Description
3-Bromopropionitrile was used as the initiator in the atom transfer radical polymerization of acrylonitrile. It was used in coupling reactions of a number of aromatic and heteroaromatic phenols with alkyl, acyl or benzoyl halides in acetonitrile with cesium fluoride-Celite.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Bromopropionitrile was used as the initiator in the atom transfer radical polymerization of acrylonitrile. It was used in coupling reactions of a number of aromatic and heteroaromatic phenols with alkyl, acyl or benzoyl halides in acetonitrile with cesium fluoride-Celite.
Solubility
Soluble in ethanol, ether.
Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
Chemical Identifiers
| 2417-90-5 | |
| 133.976 | |
| CQZIEDXCLQOOEH-UHFFFAOYSA-N | |
| 17020 | |
| C(CBr)C#N |
| C3H4BrN | |
| MFCD00001951 | |
| 3-bromopropionitrile, propanenitrile, 3-bromo, 2-bromoethyl cyanide, 3-bromopropiononitrile, 1-bromo-2-cyanoethane, beta-bromopropionitrile, 2-cyanoethyl bromide, propionitrile, 3-bromo, usaf do-51, beta-bromopriopionitrile | |
| 3-bromopropanenitrile |
Specifications
| 2417-90-5 | |
| 200°C | |
| 0.98 | |
| 1.481 | |
| 25 g | |
| 1738565 | |
| Soluble in ethanol,ether. | |
| C(CBr)C#N | |
| 133.976 | |
| 133.98 | |
| 3-Bromopropionitrile |
| 1.615 | |
| 97°C (206°F) | |
| C3H4BrN | |
| MFCD00001951 | |
| UN3276 | |
| 3-bromopropionitrile, propanenitrile, 3-bromo, 2-bromoethyl cyanide, 3-bromopropiononitrile, 1-bromo-2-cyanoethane, beta-bromopropionitrile, 2-cyanoethyl bromide, propionitrile, 3-bromo, usaf do-51, beta-bromopriopionitrile | |
| CQZIEDXCLQOOEH-UHFFFAOYSA-N | |
| 3-bromopropanenitrile | |
| 17020 | |
| 98% |
Safety and Handling
GHS H Statement
H300-H310-H330-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H301-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber : 219-332-4
RTECSNumber : UG1050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only