Learn More
3-Bromopyridine-2-carboxylic acid, 97%
CAS: 30683-23-9 | C6H4BrNO2 | 202.007 g/mol
$146.00 - $517.23
Chemical Identifiers
| CAS | 30683-23-9 |
|---|---|
| Molecular Formula | C6H4BrNO2 |
| Molecular Weight (g/mol) | 202.007 |
| MDL Number | MFCD01320380 |
| InChI Key | KBDIRPOTVAODSA-UHFFFAOYSA-N |
| Synonym | 3-bromopicolinic acid, 3-bromo-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 3-bromo, 3-bromopyridine-2-carboxylicacid, 3-bromo-2-pyridine carboxylic acid, 3-bromo-pyridine-2-carboxylic acid, pubchem5070, bromopyridine carboxylic, acmc-1cpch, 3-bromo-2-picolinic acid |
| PubChem CID | 2050129 |
| IUPAC Name | 3-bromopyridine-2-carboxylic acid |
| SMILES | C1=CC(=C(N=C1)C(=O)O)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6425806
|
Thermo Scientific Chemicals
H6425806 |
5 g |
Each for $146.00
|
|
|||||
|
AAH6425814
|
Thermo Scientific Chemicals
H6425814 |
25 g |
Each for $517.23
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 30683-23-9 | |
| 202.007 | |
| KBDIRPOTVAODSA-UHFFFAOYSA-N | |
| 2050129 | |
| C1=CC(=C(N=C1)C(=O)O)Br |
| C6H4BrNO2 | |
| MFCD01320380 | |
| 3-bromopicolinic acid, 3-bromo-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 3-bromo, 3-bromopyridine-2-carboxylicacid, 3-bromo-2-pyridine carboxylic acid, 3-bromo-pyridine-2-carboxylic acid, pubchem5070, bromopyridine carboxylic, acmc-1cpch, 3-bromo-2-picolinic acid | |
| 3-bromopyridine-2-carboxylic acid |
Specifications
| 30683-23-9 | |
| 1.813 g/mL | |
| MFCD01320380 | |
| 472171 | |
| KBDIRPOTVAODSA-UHFFFAOYSA-N | |
| 3-bromopyridine-2-carboxylic acid | |
| 2050129 | |
| 97% | |
| 3-Bromopyridine-2-carboxylic acid |
| 141°C to 144°C | |
| C6H4BrNO2 | |
| 5 g | |
| 3-bromopicolinic acid, 3-bromo-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 3-bromo, 3-bromopyridine-2-carboxylicacid, 3-bromo-2-pyridine carboxylic acid, 3-bromo-pyridine-2-carboxylic acid, pubchem5070, bromopyridine carboxylic, acmc-1cpch, 3-bromo-2-picolinic acid | |
| C1=CC(=C(N=C1)C(=O)O)Br | |
| 202.007 | |
| 202.01 | |
| Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only