missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromoquinoline, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 154270250

 View more versions of this product

Catalog No. AC154270250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Bromoquinoline
97.5
13.0°C to 15.0°C
1.5300g/mL
>112°C
97.5% min. (GC)
C9H6BrN
25 g
1.53
ZGIKWINFUGEQEO-UHFFFAOYSA-N
3-bromoquinoline
21413
98%
5332-24-1
100.0
Colorless to Yellow
274.0°C to 276.0°C
Authentic
Glass bottle
1.6630 to 1.6650
20, 363
quinoline, 3-bromo, 3-bromoquinliine, 3-bromo-quinoline, 3-bromo quinoline, 3-bromoquinolin, pubchem5814, 3-bromoquinoline, acmc-1ax3i
C1=CC=C2C(=C1)C=C(C=N2)Br
208.06
208.06
Liquid

Chemical Identifiers

5332-24-1
208.06
quinoline, 3-bromo, 3-bromoquinliine, 3-bromo-quinoline, 3-bromo quinoline, 3-bromoquinolin, pubchem5814, 3-bromoquinoline, acmc-1ax3i
3-bromoquinoline
C9H6BrN
ZGIKWINFUGEQEO-UHFFFAOYSA-N
21413
C1=CC=C2C(=C1)C=C(C=N2)Br
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-237-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only