missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromothiophene-2-carboxylic Acid 98.0+%, TCI America™
$42.87 - $130.48
Chemical Identifiers
| CAS | 7311-64-0 |
|---|---|
| Molecular Formula | C5H3BrO2S |
| Molecular Weight (g/mol) | 207.041 |
| MDL Number | MFCD00052291 |
| InChI Key | VQQLWBUTTMNMFT-UHFFFAOYSA-N |
| Synonym | 3-bromo-2-thiophenecarboxylic acid, 3-bromo-2-thenoic acid, 2-thiophenecarboxylic acid, 3-bromo, 3-bromo-thiophene-2-carboxylic acid, maybridge1_007344, 3-bromothiophene-2-carboxylicacid, bromothiophenecarboxylic, pubchem10096, ksc494c9j, bromo-thiophene-2-carboxylic acid |
| PubChem CID | 2739688 |
| IUPAC Name | 3-bromothiophene-2-carboxylic acid |
| SMILES | C1=CSC(=C1Br)C(=O)O |
Chemical Identifiers
| 7311-64-0 | |
| 207.041 | |
| VQQLWBUTTMNMFT-UHFFFAOYSA-N | |
| 2739688 | |
| C1=CSC(=C1Br)C(=O)O |
| C5H3BrO2S | |
| MFCD00052291 | |
| 3-bromo-2-thiophenecarboxylic acid, 3-bromo-2-thenoic acid, 2-thiophenecarboxylic acid, 3-bromo, 3-bromo-thiophene-2-carboxylic acid, maybridge1_007344, 3-bromothiophene-2-carboxylicacid, bromothiophenecarboxylic, pubchem10096, ksc494c9j, bromo-thiophene-2-carboxylic acid | |
| 3-bromothiophene-2-carboxylic acid |
Specifications
| 7311-64-0 | |
| White-Yellow | |
| MFCD00052291 | |
| 3-bromo-2-thiophenecarboxylic acid, 3-bromo-2-thenoic acid, 2-thiophenecarboxylic acid, 3-bromo, 3-bromo-thiophene-2-carboxylic acid, maybridge1_007344, 3-bromothiophene-2-carboxylicacid, bromothiophenecarboxylic, pubchem10096, ksc494c9j, bromo-thiophene-2-carboxylic acid | |
| C1=CSC(=C1Br)C(=O)O | |
| 207.041 | |
| 207.04 | |
| Crystalline Powder |
| 198°C | |
| C5H3BrO2S | |
| 1 g | |
| VQQLWBUTTMNMFT-UHFFFAOYSA-N | |
| 3-bromothiophene-2-carboxylic acid | |
| 2739688 | |
| ≥98.0% (GC,T) | |
| 3-Bromothiophene-2-carboxylic Acid |
Safety and Handling
TSCA : No