missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromothiophene, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1402230

 View more versions of this product

Catalog No. AAA1402230


Explore available promotions

Description

Description

3-Bromothiophene is used in the preparation of derivatives such as thienylenic alpha, μ-diformyl-alpha-oligothiophenes and 3-lithiothiophene. It is also used in the synthesis of 3-lithiothiophene by reacting with n-butyllithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromothiophene is used in the preparation of derivatives such as thienylenic alpha, µ-diformyl-alpha-oligothiophenes and 3-lithiothiophene. It is also used in the synthesis of 3-lithiothiophene by reacting with n-butyllithium.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
Specifications

Specifications

3-Bromothiophene
1.722
63°C (145°F)
C4H3BrS
MFCD00005464
UN2810
3-thienyl bromide, thiophene, 3-bromo, 3-bromo thiophene, beta-bromothiophene, .beta.-bromothiophene, 3-bromo-thiophene, bromothiophene 3-, 3-bromothiphene, 3-bromthiophene, pubchem5306
XCMISAPCWHTVNG-UHFFFAOYSA-N
3-bromothiophene
13383
97%
872-31-1
157°C to 158°C
Pungent
1.591
250 g
105338
Immiscible with water.
BrC1=CSC=C1
163.03
163.04

Chemical Identifiers

872-31-1
163.03
XCMISAPCWHTVNG-UHFFFAOYSA-N
13383
BrC1=CSC=C1
C4H3BrS
MFCD00005464
3-thienyl bromide, thiophene, 3-bromo, 3-bromo thiophene, beta-bromothiophene, .beta.-bromothiophene, 3-bromo-thiophene, bromothiophene 3-, 3-bromothiphene, 3-bromthiophene, pubchem5306
3-bromothiophene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H227-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H301-H310+H330-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: I

EINECSNumber : 212-821-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only