missing translation for 'onlineSavingsMsg'
Learn More

3-Buten-1-ol, 98+%

CAS: 627-27-0 | C4H8O | 72.107 g/mol

$73.62 - $236.55

Chemical Identifiers

CAS 627-27-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD00002959
InChI Key ZSPTYLOMNJNZNG-UHFFFAOYSA-N
Synonym 3-buten-1-ol, allylcarbinol, 3-butenyl alcohol, 1-buten-4-ol, vinylethyl alcohol, 3-butene-1-ol, ch2=chch2ch2oh, homoallyl alcohol, unii-3db2krm1i9, 1-hydroxy-3-butene
PubChem CID 69389
IUPAC Name but-3-en-1-ol
SMILES C=CCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0076706
View Documents
Thermo Scientific Chemicals
L0076706
5 g
Each for $73.62
Only null left
 
AAL0076714
View Documents
Thermo Scientific Chemicals
L0076714
25 g
Each for $236.55
Only null left
 
Description

Description

3-Buten-1-ol is employed in a study of the Mn-catalyzed hydrohydrazination of olefins. Also used in a study of the conversion of propargylic acetates to ethers catalyzed by ferric chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Buten-1-ol is employed in a study of the Mn-catalyzed hydrohydrazination of olefins. Also used in a study of the conversion of propargylic acetates to ethers catalyzed by ferric chloride.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

627-27-0
72.107
ZSPTYLOMNJNZNG-UHFFFAOYSA-N
69389
C=CCCO
C4H8O
MFCD00002959
3-buten-1-ol, allylcarbinol, 3-butenyl alcohol, 1-buten-4-ol, vinylethyl alcohol, 3-butene-1-ol, ch2=chch2ch2oh, homoallyl alcohol, unii-3db2krm1i9, 1-hydroxy-3-butene
but-3-en-1-ol

Specifications

627-27-0
113°C to 114°C
C4H8O
MFCD00002959
UN1987
3-buten-1-ol, allylcarbinol, 3-butenyl alcohol, 1-buten-4-ol, vinylethyl alcohol, 3-butene-1-ol, ch2=chch2ch2oh, homoallyl alcohol, unii-3db2krm1i9, 1-hydroxy-3-butene
ZSPTYLOMNJNZNG-UHFFFAOYSA-N
but-3-en-1-ol
69389
≥98%
0.841
32°C (89°F)
1.421
5 g
1633504
Soluble in water.
C=CCCO
72.107
72.11
3-Buten-1-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

EINECSNumber : 210-991-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only