missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Buten-1-ol, 98+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 295751000

Catalog No. AC295751000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Buten-1-ol
98.0
0.8300g/mL
32°C
98% min. (GC)
C4H8O
H2C=CHCH2CH2OH
100mL
0.83
Solubility in water: soluble in water. Other solubilities: miscible with ethanol,ether,acetone
C=CCCO
72.11
72.11
627-27-0
100.0
112.0°C to 114.0°C
Authentic
Glass bottle
1.4200 to 1.4220
MFCD00002959
01, 441
3-buten-1-ol, allylcarbinol, 3-butenyl alcohol, 1-buten-4-ol, vinylethyl alcohol, 3-butene-1-ol, ch2=chch2ch2oh, homoallyl alcohol, unii-3db2krm1i9, 1-hydroxy-3-butene
ZSPTYLOMNJNZNG-UHFFFAOYSA-N
but-3-en-1-ol
69389
98+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Warning

EINECSNumber : 210-991-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only