missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-3-Carene 90.0+%, TCI America™
$44.73 - $141.67
Chemical Identifiers
| CAS | 498-15-7 |
|---|---|
| Molecular Formula | C10H16 |
| Molecular Weight (g/mol) | 136.24 |
| MDL Number | MFCD00066417 |
| InChI Key | BQOFWKZOCNGFEC-BDAKNGLRSA-N |
| PubChem CID | 443156 |
| ChEBI | CHEBI:7 |
| IUPAC Name | (1S,6R)-3,7,7-trimethylbicyclo[4.1.0]hept-3-ene |
| SMILES | CC1=CC[C@@H]2[C@H](C1)C2(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C004725ML
|
TCI America
C004725ML |
25 mL |
Each for $44.73
|
|
|||||
|
C0047500ML
|
TCI America
C0047500ML |
500 mL |
Each for $141.67
|
|
|||||
Chemical Identifiers
| 498-15-7 | |
| 136.24 | |
| BQOFWKZOCNGFEC-BDAKNGLRSA-N | |
| CHEBI:7 | |
| CC1=CC[C@@H]2[C@H](C1)C2(C)C |
| C10H16 | |
| MFCD00066417 | |
| 443156 | |
| (1S,6R)-3,7,7-trimethylbicyclo[4.1.0]hept-3-ene |
Specifications
| 498-15-7 | |
| 170°C | |
| MFCD00066417 | |
| 2319 | |
| CC1=CC[C@@H]2[C@H](C1)C2(C)C | |
| 136.24 | |
| CHEBI:7 | |
| ≥90.0% (GC) | |
| (+)-3-Carene |
| Yellow | |
| C10H16 | |
| 25 mL | |
| BQOFWKZOCNGFEC-BDAKNGLRSA-N | |
| (1S,6R)-3,7,7-trimethylbicyclo[4.1.0]hept-3-ene | |
| 443156 | |
| 136.24 | |
| Liquid |
Safety and Handling
EINECSNumber : (4)-0589
RTECSNumber : FH8400000
TSCA : Yes