Learn More
3-Chloro-2,5,6-trifluoropyridine, 98+%
CAS: 2879-42-7 | C5HClF3N | 167.52 g/mol
Supplier: Thermo Scientific Chemicals L1954314
Description
3-Chloro-2,5,6-trifluoropyridine is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Chloro-2,5,6-trifluoropyridine is used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix in water.
Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
| 3-Chloro-2,5,6-trifluoropyridine | |
| 132°C to 134°C | |
| Unpleasant | |
| 1.456 | |
| 25 g | |
| 1424722 | |
| Not miscible or difficult to mix in water. | |
| FC1=CC(Cl)=C(F)N=C1F | |
| 167.52 | |
| 167.52 |
| 2879-42-7 | |
| >100°C (212°F) | |
| C5HClF3N | |
| MFCD03001160 | |
| UN1760 | |
| pyridine,3-chloro-2,5,6-trifluoro, pyridine, 3-chloro-2,5,6-trifluoro, 3-chloro-2,5,6-trifluoro-pyridine, acmc-1cekn, 2,3,6-trifluoro-5-chloropyridine, 5-chloro-2,3,6-trifluoropyridine, 3-chloranyl-2,5,6-tris fluoranyl pyridine | |
| OROKHXKJSDPRET-UHFFFAOYSA-N | |
| 3-chloro-2,5,6-trifluoropyridine | |
| 2783252 | |
| ≥98% |
Chemical Identifiers
| 2879-42-7 | |
| 167.52 | |
| OROKHXKJSDPRET-UHFFFAOYSA-N | |
| 3-chloro-2,5,6-trifluoropyridine |
| C5HClF3N | |
| MFCD03001160 | |
| 2783252 | |
| FC1=CC(Cl)=C(F)N=C1F |
Safety and Handling
GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only