missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-2,6-dimethylaniline, 99%, Thermo Scientific Chemicals

$107.58 - $1382.41

Chemical Identifiers

CAS 26829-77-6
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD02683086
InChI Key FGMXFOTYCHZCLA-UHFFFAOYSA-N
Synonym 3-chloro-2,6-dimethyl-phenylamine, acmc-20an7h, 3-chloro-2,6-xylidine, 3-chloro-2,6-dimethylphenylamine, 3-chloranyl-2,6-dimethyl-aniline, benzenamine,3-chloro-2,6-dimethyl
PubChem CID 7023016
IUPAC Name 3-chloro-2,6-dimethylaniline
SMILES CC1=C(C(=C(C=C1)Cl)C)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2223209
View Documents
Thermo Scientific Chemicals
B2223209
10 g
Each for $107.58
 
AAB2223218
View Documents
Thermo Scientific Chemicals
B2223218
50 g
Each for $357.93
 
AAB2223230
View Documents
Thermo Scientific Chemicals
B2223230
250 g
Each for $1,382.41
 
Description

Description

3-cyano- 2,4-dimethylchlorobenzene was obtained from 3-Chloro-2,6-dimethylaniline which upon hydrolysis gives acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-cyano- 2,4-dimethylchlorobenzene was obtained from 3-Chloro-2,6-dimethylaniline which upon hydrolysis gives acid.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

26829-77-6
155.625
FGMXFOTYCHZCLA-UHFFFAOYSA-N
7023016
CC1=C(C(=C(C=C1)Cl)C)N
C8H10ClN
MFCD02683086
3-chloro-2,6-dimethyl-phenylamine, acmc-20an7h, 3-chloro-2,6-xylidine, 3-chloro-2,6-dimethylphenylamine, 3-chloranyl-2,6-dimethyl-aniline, benzenamine,3-chloro-2,6-dimethyl
3-chloro-2,6-dimethylaniline

Specifications

30°C to 32°C
136°C to 138°C (17 mmHg)
C8H10ClN
3-chloro-2,6-dimethyl-phenylamine, acmc-20an7h, 3-chloro-2,6-xylidine, 3-chloro-2,6-dimethylphenylamine, 3-chloranyl-2,6-dimethyl-aniline, benzenamine,3-chloro-2,6-dimethyl
FGMXFOTYCHZCLA-UHFFFAOYSA-N
3-chloro-2,6-dimethylaniline
7023016
99%
26829-77-6
10 g
MFCD02683086
Slightly soluble in water.
CC1=C(C(=C(C=C1)Cl)C)N
155.625
155.63
3-Chloro-2,6-dimethylaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only