Learn More
3-Chloro-2-butanone, 96%, Thermo Scientific Chemicals
$80.40 - $247.14
Chemical Identifiers
CAS | 4091-39-8 |
---|---|
Molecular Formula | C4H7ClO |
Molecular Weight (g/mol) | 106.549 |
MDL Number | MFCD00000865 |
InChI Key | OIMRLHCSLQUXLL-UHFFFAOYSA-N |
Synonym | 3-chloro-2-butanone, 2-butanone, 3-chloro, 3-chlorobutanone, 1-acetyl-1-chloroethane, 1-chloroethyl methyl ketone, 2-chloro-3-butanone, 2-butanone, 3-chloro-, +, chloro-mek, 3-chloro-2-butanon, 3-chlorobutan-2-on |
PubChem CID | 20026 |
IUPAC Name | 3-chlorobutan-2-one |
SMILES | CC(C(=O)C)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1415822
|
Thermo Scientific Chemicals
L1415822 |
100 g |
Each for $80.40
|
|
|||||
AAL1415836
|
Thermo Scientific Chemicals
L1415836 |
500 g |
Each for $247.14
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4091-39-8 | |
106.549 | |
OIMRLHCSLQUXLL-UHFFFAOYSA-N | |
20026 | |
CC(C(=O)C)Cl |
C4H7ClO | |
MFCD00000865 | |
3-chloro-2-butanone, 2-butanone, 3-chloro, 3-chlorobutanone, 1-acetyl-1-chloroethane, 1-chloroethyl methyl ketone, 2-chloro-3-butanone, 2-butanone, 3-chloro-, +, chloro-mek, 3-chloro-2-butanon, 3-chlorobutan-2-on | |
3-chlorobutan-2-one |
Specifications
4091-39-8 | |
1.055 | |
28°C (82°F) | |
1.421 | |
100 g | |
385637 | |
OIMRLHCSLQUXLL-UHFFFAOYSA-N | |
3-chlorobutan-2-one | |
20026 | |
96% |
<-60°C | |
113°C to 116°C | |
C4H7ClO | |
MFCD00000865 | |
UN1224 | |
3-chloro-2-butanone, 2-butanone, 3-chloro, 3-chlorobutanone, 1-acetyl-1-chloroethane, 1-chloroethyl methyl ketone, 2-chloro-3-butanone, 2-butanone, 3-chloro-, +, chloro-mek, 3-chloro-2-butanon, 3-chlorobutan-2-on | |
CC(C(=O)C)Cl | |
106.549 | |
106.55 | |
3-Chloro-2-butanone |
Safety and Handling
GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber : 223-834-9
RTECSNumber : EL7062000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only